This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00937996_m1
See other PHACTR3 GE Assays ›
Gene Symbol:
PHACTR3
Gene Name:
phosphatase and actin regulator 3
Gene Aliases:
C20orf101, H17739, PPP1R123, SCAPIN1, SCAPININ
Chromosome Location:
Chr.20: 59577497 - 59849829 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00937996_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHACTR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199505.1 NP_001186434.1 11 - 12 1582 94
NM_001199506.1 NP_001186435.1 11 - 12 1577 94
NM_001281507.1 NP_001268436.1 11 - 12 1681 94
NM_080672.4 NP_542403.1 11 - 12 2059 94
NM_183244.1 NP_899067.1 11 - 12 1702 94
NM_183246.1 NP_899069.1 10 - 11 1492 94
XM_011528525.2 XP_011526827.1 11 - 12 1688 94
XM_011528526.2 XP_011526828.1 10 - 11 1494 94
XM_017027626.1 XP_016883115.1 11 - 12 1791 94
XM_017027627.1 XP_016883116.1 11 - 12 1736 94
XM_017027628.1 XP_016883117.1 10 - 11 1384 94
XM_017027629.1 XP_016883118.1 10 - 11 1527 94
XM_017027630.1 XP_016883119.1 10 - 11 1355 94
XM_017027631.1 XP_016883120.1 11 - 12 1618 94
GenBank mRNA AB098521.1 - 11 - 12 1702 94
AB098522.1 - 10 - 11 1492 94
AJ311122.2 - 11 - 12 2058 94
AJ617581.1 - 11 - 12 1577 94
AK091632.1 - 11 - 12 1704 94
AK098788.1 - 11 - 12 1582 94
AK295459.1 - 11 - 12 1681 94
AK314493.1 - 11 - 12 1789 94
AK316047.1 - 11 - 12 1574 94
BC108303.1 - 11 - 12 2059 4179728 94
BC117362.1 - 11 - 12 1947 40125913 94
BC117364.1 - 11 - 12 1950 40125915 94

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PHACTR3

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of catalytic activity
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0