This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00938001_m1
See other PHACTR3 GE Assays ›
Gene Symbol:
PHACTR3
Gene Name:
phosphatase and actin regulator 3
Gene Aliases:
C20orf101, H17739, PPP1R123, SCAPIN1, SCAPININ
Chromosome Location:
Chr.20: 59577497 - 59849829 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00938001_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PHACTR3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001199505.1 NP_001186434.1 4 - 5 536 60
NM_001199506.1 NP_001186435.1 4 - 5 531 60
NM_001281507.1 NP_001268436.1 4 - 5 635 60
NM_080672.4 NP_542403.1 4 - 5 1013 60
NM_183244.1 NP_899067.1 4 - 5 656 60
XM_011528525.2 XP_011526827.1 4 - 5 642 60
XM_017027626.1 XP_016883115.1 4 - 5 745 60
XM_017027627.1 XP_016883116.1 4 - 5 690 60
XM_017027631.1 XP_016883120.1 4 - 5 572 60
GenBank mRNA AB098521.1 - 4 - 5 656 60
AJ311122.2 - 4 - 5 1012 60
AJ617581.1 - 4 - 5 531 60
AK091632.1 - 4 - 5 658 60
AK098788.1 - 4 - 5 536 60
AK314493.1 - 4 - 5 743 60
AK316047.1 - 4 - 5 528 60
BC108303.1 - 4 - 5 1013 4179728 60
BC117362.1 - 4 - 5 901 40125913 60
BC117364.1 - 4 - 5 904 40125915 60

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PHACTR3

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of catalytic activity
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0