This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00938136_m1
See other ADGRL2 GE Assays ›
Gene Symbol:
ADGRL2
Gene Name:
adhesion G protein-coupled receptor L2
Gene Aliases:
CIRL2, CL2, LEC1, LPHH1, LPHN2
Chromosome Location:
Chr.1: 81306155 - 81992738 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00938136_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001297705.1 NP_001284634.1 20 - 21 3730 75
GenBank mRNA AF104266.1 - 20 - 21 3730 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100008845.hg.1

Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

G-protein coupled receptor

Gene Ontology Categories:

Function(s) Process(es)

cell surface receptor signaling pathway
G-protein coupled receptor signaling pathway
G-protein coupled receptor activity
latrotoxin receptor activity
carbohydrate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-55ff9ddd88-g6tbm:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0