This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00938193_m1
See other DLG1 GE Assays ›
Gene Symbol:
DLG1
Gene Name:
discs large MAGUK scaffold protein 1
Gene Aliases:
DLGH1, SAP-97, SAP97, dJ1061C18.1.1, hdlg
Chromosome Location:
Chr.3: 197042560 - 197299272 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00938193_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DLG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098424.1 NP_001091894.1 15 - 16 1831 121
NM_001204386.1 NP_001191315.1 14 - 15 1732 121
NM_001204387.1 NP_001191316.1 10 - 11 1616 121
NM_001204388.1 NP_001191317.1 10 - 11 1616 121
NM_001290983.1 NP_001277912.1 15 - 16 1900 121
NM_004087.2 NP_004078.2 15 - 16 1831 121
XM_005269289.3 XP_005269346.1 15 - 16 2113 121
XM_011512502.2 XP_011510804.1 15 - 16 2113 121
XM_011512503.1 XP_011510805.1 14 - 15 2011 121
XM_011512505.1 XP_011510807.1 14 - 15 2011 121
XM_011512506.1 XP_011510808.1 13 - 14 1957 121
XM_011512509.1 XP_011510811.1 14 - 15 2011 121
XM_017005800.1 XP_016861289.1 15 - 16 2248 121
XM_017005801.1 XP_016861290.1 15 - 16 1768 121
XM_017005802.1 XP_016861291.1 15 - 16 1838 121
XM_017005803.1 XP_016861292.1 15 - 16 1759 121
XM_017005804.1 XP_016861293.1 15 - 16 2113 121
XM_017005805.1 XP_016861294.1 15 - 16 1930 121
XM_017005806.1 XP_016861295.1 14 - 15 2011 121
XM_017005807.1 XP_016861296.1 14 - 15 2110 121
XM_017005808.1 XP_016861297.1 14 - 15 1669 121
XM_017005809.1 XP_016861298.1 14 - 15 1771 121
XM_017005810.1 XP_016861299.1 14 - 15 1669 121
XM_017005811.1 XP_016861300.1 13 - 14 1957 121
XM_017005812.1 XP_016861301.1 13 - 14 1611 121
XM_017005813.1 XP_016861302.1 13 - 14 1615 121
XM_017005814.1 XP_016861303.1 13 - 14 1718 121
XM_017005815.1 XP_016861304.1 13 - 14 1957 121
XM_017005816.1 XP_016861305.1 14 - 15 2110 121
XM_017005817.1 XP_016861306.1 14 - 15 1641 121
XM_017005818.1 XP_016861307.1 14 - 15 1743 121
XM_017005819.1 XP_016861308.1 14 - 15 1660 121
XM_017005820.1 XP_016861309.1 13 - 14 1675 121
XM_017005821.1 XP_016861310.1 11 - 12 4116 121
XM_017005822.1 XP_016861311.1 11 - 12 4118 121
XM_017005823.1 XP_016861312.1 10 - 11 4013 121
XM_017005824.1 XP_016861313.1 5 - 6 632 121
GenBank mRNA AB209536.2 - 7 - 8 954 121
AK293967.1 - 10 - 11 1335 121
AK294772.1 - 10 - 11 1343 121
AK294855.1 - 10 - 11 1616 121
AK310596.1 - 14 - 15 1670 121
AK310893.1 - 15 - 16 2115 121
BC140841.1 - 15 - 16 1796 9021709 121
BC144651.1 - 14 - 15 1697 9053182 121
EF553524.1 - 15 - 16 1900 121
U13896.1 - 15 - 16 1829 121
U13897.1 - 15 - 16 1829 121

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300013736.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03002174.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DLG1

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
branching involved in ureteric bud morphogenesis
immunological synapse formation
endothelial cell proliferation
lens development in camera-type eye
T cell cytokine production
protein dephosphorylation
actin filament organization
mitotic cell cycle checkpoint
establishment or maintenance of cell polarity
chemical synaptic transmission
nervous system development
positive regulation of cell proliferation
regulation of cell shape
viral process
single organismal cell-cell adhesion
peristalsis
positive regulation of actin filament polymerization
cortical actin cytoskeleton organization
astral microtubule organization
membrane raft organization
regulation of myelination
activation of protein kinase activity
ion transmembrane transport
cellular protein complex localization
T cell activation
negative regulation of T cell proliferation
regulation of membrane potential
amyloid precursor protein metabolic process
receptor clustering
positive regulation of potassium ion transport
cortical microtubule organization
establishment or maintenance of epithelial cell apical/basal polarity
negative regulation of mitotic cell cycle
GMP metabolic process
GDP metabolic process
reproductive structure development
embryonic skeletal system morphogenesis
smooth muscle tissue development
negative regulation of epithelial cell proliferation
establishment of centrosome localization
negative regulation of protein kinase B signaling
hard palate development
negative regulation of ERK1 and ERK2 cascade
bicellular tight junction assembly
protein localization to plasma membrane
positive regulation of establishment of protein localization to plasma membrane
receptor localization to synapse
regulation of NIK/NF-kappaB signaling
regulation of sodium ion transmembrane transport
negative regulation of p38MAPK cascade
guanylate kinase activity
phosphoprotein phosphatase activity
protein binding
protein C-terminus binding
cytoskeletal protein binding
ligand-gated ion channel activity
potassium channel regulator activity
protein kinase binding
phosphatase binding
mitogen-activated protein kinase kinase binding
protein complex scaffold
ionotropic glutamate receptor binding
ion channel binding
L27 domain binding
cadherin binding involved in cell-cell adhesion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 TC03002174.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 ENST00000314062.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 ENST00000346964.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 ENST00000357674.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 ENST00000359922.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 ENST00000381807.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 ENST00000392382.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 ENST00000419354.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 ENST00000422288.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 ENST00000443183.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 ENST00000448528.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 ENST00000450955.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 ENST00000452595.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 NM_001098424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 NM_001204386.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 NM_001204387.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 NM_001204388.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03038426.hg.1 NM_004087.2 Refseq
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000346964.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000357674.8 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000392381.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000392382.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000419227.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000419354.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000422288.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000443183.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000448528.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000450955.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000452595.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000471733.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 NM_001098424.1 Refseq
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 NM_001204386.1 Refseq
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 NM_001204387.1 Refseq
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 NM_001204388.1 Refseq
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 NM_001290983.1 Refseq
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 NM_004087.2 Refseq
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000346964.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000357674.8 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000392381.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000392382.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000419227.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000419354.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000422288.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000443183.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000448528.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000450955.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000452595.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 ENST00000471733.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 NM_001098424.1 Refseq
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 NM_001204386.1 Refseq
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 NM_001204387.1 Refseq
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 NM_001204388.1 Refseq
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 NM_001290983.1 Refseq
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 NM_004087.2 Refseq
Affymetrix Human Clariom D Assay PSR0300202690.hg.1 TC0300013736.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0