This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00938383_g1
See other EHMT2 GE Assays ›
Gene Symbol:
EHMT2
Gene Name:
euchromatic histone lysine methyltransferase 2
Gene Aliases:
BAT8, C6orf30, G9A, GAT8, KMT1C, NG36
Chromosome Location:
Chr.6: 31879759 - 31897707 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00938383_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EHMT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001289413.1 NP_001276342.1 24 - 25 3352 97
NM_001318833.1 NP_001305762.1 26 - 27 3559 97
NM_006709.4 NP_006700.3 26 - 27 3309 97
NM_025256.6 NP_079532.5 25 - 26 3207 97
XM_005248824.3 XP_005248881.1 25 - 26 3450 97
XM_006714974.1 XP_006715037.1 25 - 26 3475 97
XM_006714975.1 XP_006715038.1 24 - 25 3373 97
XM_006714976.1 XP_006715039.1 26 - 27 3313 97
XM_017010202.1 XP_016865691.1 25 - 26 3211 97
GenBank mRNA AB209433.2 - 25 - 26 3392 97
AJ315532.1 - 26 - 27 3279 97
AK092866.1 - 9 - 10 2224 97
AK302904.1 - 24 - 25 3352 97
BC002686.1 - 23 - 24 2779 97
BC009351.1 - 26 - 27 3276 97
BC018718.1 - 26 - 27 3267 4123580 97
BC020970.1 - 25 - 26 3244 97
X69838.1 - 22 - 23 2699 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600011465.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001550.hg.1, TC6_apd_hap1000091.hg.1, TC6_cox_hap2000177.hg.1, TC6_dbb_hap3000166.hg.1, TC6_mcf_hap5000152.hg.1, TC6_qbl_hap6000167.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EHMT2

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of DNA replication
DNA methylation
cellular response to starvation
histone methylation
peptidyl-lysine dimethylation
histone lysine methylation
histone H3-K9 methylation
histone H3-K27 methylation
regulation of signal transduction by p53 class mediator
p53 binding
protein binding
zinc ion binding
protein-lysine N-methyltransferase activity
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H3-K9 specific)
histone methyltransferase activity (H3-K27 specific)
C2H2 zinc finger domain binding
promoter-specific chromatin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline