This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00938598_m1
See other DHRS12 GE Assays ›
Gene Symbol:
DHRS12
Gene Name:
dehydrogenase/reductase 12
Gene Aliases:
SDR40C1
Chromosome Location:
Chr.13: 51754856 - 51804206 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00938598_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DHRS12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270424.1 NP_001257353.1 2 - 3 245 77
XM_005266528.4 XP_005266585.1 2 - 3 289 77
XM_011535235.2 XP_011533537.1 2 - 3 289 77
XM_011535236.2 XP_011533538.1 2 - 3 289 77
XM_017020749.1 XP_016876238.1 2 - 3 289 77
XM_017020750.1 XP_016876239.1 2 - 3 289 77
GenBank mRNA BC026024.1 - 2 - 3 178 77
DB481446.1 - 2 - 3 245 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1300008997.hg.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
oxidoreductase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000461948.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000461948.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000461948.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000461948.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000469957.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000469957.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000469957.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000469957.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000469957.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000472372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000472372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000472372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000472372.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000489680.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000489680.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000489680.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000489680.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000489680.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000497151.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000497151.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000497151.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000497151.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 ENST00000497151.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000218981.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000280056.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000461948.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000461948.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000461948.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000461948.2 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000469957.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000469957.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000469957.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000469957.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000469957.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000472372.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000472372.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000472372.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000472372.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000489680.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000489680.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000489680.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000489680.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000489680.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000490949.5 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000497151.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000497151.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000497151.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000497151.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 ENST00000497151.1 Ensembl
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001031719.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_001270424.1 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay TC1300008997.hg.1 NM_024705.2 Refseq
Affymetrix Human Clariom D Assay PSR1300160719.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300160719.hg.1 ENST00000497151.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300160719.hg.1 NM_001270424.1 Refseq
Affymetrix Human Clariom D Assay PSR1300160719.hg.1 ENST00000444610.6 Ensembl
Affymetrix Human Clariom D Assay PSR1300160719.hg.1 ENST00000497151.1 Ensembl
Affymetrix Human Clariom D Assay PSR1300160719.hg.1 NM_001270424.1 Refseq
Affymetrix Human Clariom D Assay PSR1300160719.hg.1 TC1300008997.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0