This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00938904_g1
See other CIAPIN1 GE Assays ›
Gene Symbol:
CIAPIN1
Gene Name:
cytokine induced apoptosis inhibitor 1
Gene Aliases:
Anamorsin, DRE2, PRO0915
Chromosome Location:
Chr.16: 57428169 - 57447528 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00938904_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CIAPIN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308347.1 NP_001295276.1 6 - 7 833 62
NM_020313.3 NP_064709.2 6 - 7 872 62
GenBank mRNA AF116609.1 - 4 - 5 483 62
AF248964.1 - 6 - 7 776 62
AK292281.1 - 6 - 7 676 62
AL136613.1 - 6 - 7 694 62
BC002568.2 - 6 - 7 714 3139226 62
BC024196.2 - 6 - 7 753 4816594 62
BC067303.1 - 6 - 7 872 30389410 62
BC071740.1 - 6 - 7 747 5563094 62
CR533545.1 - 5 - 6 591 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF116609.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CIAPIN1

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
iron-sulfur cluster assembly
hemopoiesis
methylation
negative regulation of apoptotic process
protein binding
methyltransferase activity
electron carrier activity
metal ion binding
2 iron, 2 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-4gzsq:80/100.66.74.9:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0