This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00938959_m1
See other SNAP25 GE Assays ›
Gene Symbol:
SNAP25
Gene Name:
synaptosome associated protein 25
Gene Aliases:
CMS18, RIC-4, RIC4, SEC9, SNAP, SNAP-25, SUP, bA416N4.2, dJ1068F16.2
Chromosome Location:
Chr.20: 10218694 - 10307420 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00938959_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SNAP25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001322902.1 NP_001309831.1 4 - 5 444 80
NM_003081.4 NP_003072.2 4 - 5 413 80
XM_017028021.1 XP_016883510.1 5 - 6 343 80
XM_017028022.1 XP_016883511.1 5 - 6 435 80
GenBank mRNA AK094560.1 - 4 - 5 381 80
AK098767.1 - 4 - 5 283 80
AK289647.1 - 6 - 7 574 80
BC010647.1 - 4 - 5 302 3867544 80
BT019684.1 - 3 - 4 170 80
L19760.1 - 4 - 5 258 80

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SNAP25

Panther Classification:

Molecular Function -

SNARE protein

Gene Ontology Categories:

Function(s) Process(es)

neurotransmitter uptake
chemical synaptic transmission
neurotransmitter secretion
locomotory behavior
associative learning
regulation of neuron projection development
glutamate secretion
synaptic vesicle exocytosis
synaptic vesicle docking
synaptic vesicle priming
synaptic vesicle fusion to presynaptic active zone membrane
regulation of insulin secretion
long-term synaptic potentiation
SNAP receptor activity
protein binding
syntaxin-1 binding
calcium-dependent protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000430336.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000430336.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000430336.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000430336.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000430336.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000430336.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000430336.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000492814.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000492814.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000492814.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000495883.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000495883.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000495883.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000495883.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 ENST00000495883.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000254976.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000304886.6 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000430336.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000430336.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000430336.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000430336.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000430336.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000430336.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000430336.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000492814.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000492814.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000492814.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000495883.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000495883.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000495883.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000495883.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 ENST00000495883.1 Ensembl
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_003081.4 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq
Affymetrix Human Clariom D Assay TC2000006688.hg.1 NM_130811.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-7k7vz:80/100.66.73.243:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0