This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00939222_g1
See other UNC119 GE Assays ›
Gene Symbol:
UNC119
Gene Name:
unc-119 lipid binding chaperone
Gene Aliases:
HRG4, IMD13, POC7, POC7A
Chromosome Location:
Chr.17: 28546707 - 28552633 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00939222_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UNC119
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017025289.1 XP_016880778.1 1 - 2 291 93

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017025289.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
chemical synaptic transmission
visual perception
phototransduction
protein transport
lipoprotein transport
positive regulation of protein tyrosine kinase activity
negative regulation of clathrin-mediated endocytosis
negative regulation of caveolin-mediated endocytosis
protein binding
lipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000444148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000444148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000444148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000444148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000444148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000444148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000444148.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000487844.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000487844.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000487844.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000578434.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000578434.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000578434.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000578434.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000581945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000581945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000581945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 ENST00000581945.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000301032.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000335765.8 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000444148.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000444148.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000444148.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000444148.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000444148.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000444148.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000444148.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000470125.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000484980.5 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000487844.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000487844.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000487844.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000578434.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000578434.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000578434.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000578434.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000581945.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000581945.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000581945.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 ENST00000581945.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_005148.3 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_054035.2 Refseq
Affymetrix Human Clariom D Assay TC1700010198.hg.1 NM_054035.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0