This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00940890_m1
See other NABP2 GE Assays ›
Gene Symbol:
NABP2
Gene Name:
nucleic acid binding protein 2
Gene Aliases:
OBFC2B, SOSS-B1, SSB1, hSSB1
Chromosome Location:
Chr.12: 56222039 - 56229858 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00940890_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NABP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005269148.4 XP_005269205.1 1 - 2 95 89

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005269148.4 transcripts.

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

double-strand break repair via homologous recombination
DNA repair
cellular response to DNA damage stimulus
mitotic cell cycle checkpoint
response to ionizing radiation
snRNA transcription from RNA polymerase II promoter
regulation of telomerase activity
establishment of protein localization to telomere
positive regulation of telomere capping
single-stranded DNA binding
protein binding
enzyme binding
C-rich strand telomeric DNA binding
G-rich strand telomeric DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000399713.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000399713.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000399713.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000399713.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000399713.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000399713.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000447747.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000447747.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000447747.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000447747.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000447747.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000447747.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000447747.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0400010709.hg.1 ENST00000508433.1 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000267023.8 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000341463.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000380198.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000399713.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000399713.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000399713.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000399713.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000399713.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000399713.6 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000447747.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000447747.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000447747.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000447747.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000447747.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000447747.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000447747.5 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 ENST00000479016.2 Ensembl
Affymetrix Human Clariom D Assay TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Clariom D Assay TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Clariom D Assay TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Clariom D Assay TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Clariom D Assay TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Clariom D Assay TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Clariom D Assay TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Clariom D Assay TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Clariom D Assay TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Clariom D Assay TC1200007832.hg.1 NM_024068.3 Refseq
Affymetrix Human Clariom D Assay TC0400010709.hg.1 ENST00000508433.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z4mpm:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0