This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00941089_m1
See other PTPN1 GE Assays ›
Gene Symbol:
PTPN1
Gene Name:
protein tyrosine phosphatase, non-receptor type 1
Gene Aliases:
PTP1B
Chromosome Location:
Chr.20: 50510321 - 50584762 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00941089_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPN1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278618.1 NP_001265547.1 3 - 4 471 160
NM_002827.3 NP_002818.1 4 - 5 562 160
GenBank mRNA AK290638.1 - 4 - 5 529 160
AK299830.1 - 3 - 4 409 160
AK316563.1 - 4 - 5 504 160
AU117677.1 - 4 - 5 529 160
BC015660.2 - 4 - 5 491 4844022 160
BC018164.1 - 4 - 5 472 4338750 160
BT006752.1 - 4 - 5 355 160
M31724.1 - 4 - 5 445 160
M33689.1 - 4 - 5 427 160

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000007695.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000403.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PTPN1

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
activation of signaling protein activity involved in unfolded protein response
activation of JUN kinase activity
insulin receptor signaling pathway
regulation of signal transduction
negative regulation of signal transduction
regulation of endocytosis
negative regulation of vascular endothelial growth factor receptor signaling pathway
endoplasmic reticulum unfolded protein response
actin cytoskeleton reorganization
regulation of intracellular protein transport
peptidyl-tyrosine dephosphorylation
platelet-derived growth factor receptor-beta signaling pathway
IRE1-mediated unfolded protein response
negative regulation of MAP kinase activity
negative regulation of insulin receptor signaling pathway
regulation of type I interferon-mediated signaling pathway
JAK-STAT cascade involved in growth hormone signaling pathway
positive regulation of protein tyrosine kinase activity
negative regulation of ERK1 and ERK2 cascade
cell-cell adhesion
regulation of hepatocyte growth factor receptor signaling pathway
negative regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
positive regulation of IRE1-mediated unfolded protein response
negative regulation of PERK-mediated unfolded protein response
peptidyl-tyrosine dephosphorylation involved in inactivation of protein kinase activity
positive regulation of receptor catabolic process
protein tyrosine phosphatase activity
insulin receptor binding
protein binding
zinc ion binding
enzyme binding
protein kinase binding
receptor tyrosine kinase binding
poly(A) RNA binding
ephrin receptor binding
protein phosphatase 2A binding
cadherin binding involved in cell-cell adhesion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20005742.hg.1 ENST00000371621.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20005742.hg.1 ENST00000541713.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20005742.hg.1 NM_002827.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20005742.hg.1 TC20000403.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR2000153655.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000153655.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000153655.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay PSR2000153655.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay PSR2000153655.hg.1 TC2000007695.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000153655.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay PSR2000153655.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000153655.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay PSR2000153655.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000371621.4 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 ENST00000541713.5 Ensembl
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_001278618.1 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq
Affymetrix Human Clariom D Assay TC2000007695.hg.1 NM_002827.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0