This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00941286_g1
See other POLR2C GE Assays ›
Gene Symbol:
POLR2C
Gene Name:
RNA polymerase II subunit C
Gene Aliases:
RPB3, RPB31, hRPB33, hsRPB3
Chromosome Location:
Chr.16: 57462639 - 57472009 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00941286_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR2C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032940.2 NP_116558.1 7 - 8 693 65
GenBank mRNA AF008443.1 - 7 - 8 607 65
AK293112.1 - 7 - 8 651 65
BC000409.2 - 7 - 8 624 2960943 65
BC003159.1 - 7 - 8 665 2957867 65
BC028157.1 - 7 - 8 649 5240953 65
CR542041.1 - 7 - 8 607 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600011402.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000491.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POLR2C

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
transcription-coupled nucleotide-excision repair
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
7-methylguanosine mRNA capping
fibroblast growth factor receptor signaling pathway
gene expression
gene silencing by RNA
somatic stem cell population maintenance
snRNA transcription from RNA polymerase II promoter
positive regulation of viral transcription
RNA polymerase II activity
DNA binding
protein binding
protein dimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16006834.hg.1 ENST00000219252.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16006834.hg.1 NM_032940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16006834.hg.1 TC16000491.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000563589.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000564626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000564626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000564626.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562599.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000562953.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563115.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563589.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563589.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563589.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563589.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000563589.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000564626.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000564626.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000564626.1 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000564651.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay TC1600011402.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay PSR1600190728.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay PSR1600190728.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600190728.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay PSR1600190728.hg.1 ENST00000219252.9 Ensembl
Affymetrix Human Clariom D Assay PSR1600190728.hg.1 ENST00000567982.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600190728.hg.1 NM_032940.2 Refseq
Affymetrix Human Clariom D Assay PSR1600190728.hg.1 TC1600011402.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0