This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00942566_m1
See other NDUFS4 GE Assays ›
Gene Symbol:
NDUFS4
Gene Name:
NADH:ubiquinone oxidoreductase subunit S4
Gene Aliases:
AQDQ, CI-18, CI-18 kDa, CI-AQDQ
Chromosome Location:
Chr.5: 53560610 - 53683341 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00942566_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NDUFS4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001318051.1 NP_001304980.1 2 - 3 206 124
NM_002495.3 NP_002486.1 2 - 3 206 124
NR_134475.1 3 - 4 330 124
GenBank mRNA AF020351.1 - 2 - 3 184 124
AK307973.1 - 2 - 3 184 124
BC005270.1 - 2 - 3 200 3681312 124
BU145342.1 - 2 - 3 180 124
BU954271.1 - 2 - 3 198 124
CD692133.1 - 3 - 4 346 124
DB489661.1 - 2 - 3 205 124
KU178226.1 - 2 - 3 176 124

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500007378.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000221.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NDUFS4

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein phosphorylation
mitochondrial electron transport, NADH to ubiquinone
brain development
cAMP-mediated signaling
mitochondrial respiratory chain complex I assembly
cellular respiration
positive regulation of fibroblast proliferation
response to cAMP
reactive oxygen species metabolic process
NADH dehydrogenase (ubiquinone) activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05002754.hg.1 TC05000221.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05002754.hg.1 ENST00000296684.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05002754.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Clariom D Assay TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay TC0500007378.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay PSR0500149901.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay PSR0500149901.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149901.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500149901.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149901.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149901.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500149901.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay PSR0500149901.hg.1 ENST00000296684.9 Ensembl
Affymetrix Human Clariom D Assay PSR0500149901.hg.1 ENST00000502423.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149901.hg.1 ENST00000506765.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500149901.hg.1 ENST00000506974.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149901.hg.1 ENST00000507026.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500149901.hg.1 ENST00000509443.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500149901.hg.1 NM_002495.3 Refseq
Affymetrix Human Clariom D Assay PSR0500149901.hg.1 TC0500007378.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0