This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00943405_g1
See other LIPE GE Assays ›
Gene Symbol:
LIPE
Gene Name:
lipase E, hormone sensitive type
Gene Aliases:
AOMS4, FPLD6, HSL, LHS
Chromosome Location:
Chr.19: 42401507 - 42427426 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00943405_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LIPE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005357.3 NP_005348.2 3 - 4 1784 86
XM_005258937.3 XP_005258994.1 3 - 4 1779 86
XM_005258938.4 XP_005258995.1 3 - 4 770 86
XM_005258939.3 XP_005258996.2 4 - 5 808 86
XM_005258940.3 XP_005258997.1 3 - 4 709 86
XM_006723218.2 XP_006723281.1 3 - 4 909 86
XM_017026810.1 XP_016882299.1 3 - 4 649 86
GenBank mRNA AK292482.1 - 3 - 4 1764 86
BC029301.1 - 3 - 4 1329 86
BC029961.1 - 3 - 4 1771 86
U40002.1 - 3 - 4 1784 86

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900010802.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19001573.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LIPE

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
cholesterol metabolic process
lipid catabolic process
triglyceride catabolic process
long-chain fatty acid catabolic process
diacylglycerol catabolic process
triglyceride lipase activity
protein binding
protein kinase binding
hormone-sensitive lipase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19021447.hg.1 ENST00000244289.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19021447.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19021447.hg.1 TC19001573.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000597001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000597001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000597001.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000597620.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000597620.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000597620.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000597620.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000597620.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000597620.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000599211.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000599211.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000599211.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000599211.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000599211.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000599783.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000599783.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000599783.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000599783.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000599783.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000599918.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000599918.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000599918.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000599918.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000599918.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000600224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000600224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000600224.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000601189.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000601189.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay PSR1900189406.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900189406.hg.1 ENST00000599211.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900189406.hg.1 ENST00000599783.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900189406.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900189406.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay PSR1900189406.hg.1 TC1900010802.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900189406.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900189406.hg.1 ENST00000599211.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900189406.hg.1 ENST00000599783.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900189406.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900189406.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay PSR1900189404.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900189404.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900189404.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay PSR1900189404.hg.1 TC1900010802.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900189404.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900189404.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900189404.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000244289.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000597001.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000597001.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000597001.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000597620.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000597620.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000597620.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000597620.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000597620.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000597620.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000599211.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000599211.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000599211.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000599211.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000599211.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000599783.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000599783.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000599783.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000599783.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000599783.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000599918.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000599918.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000599918.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000599918.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000599918.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000600224.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000600224.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000600224.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000601189.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000601189.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 ENST00000602000.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq
Affymetrix Human Clariom D Assay TC1900010802.hg.1 NM_005357.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0