This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00944504_g1
See other TUFM GE Assays ›
Gene Symbol:
TUFM
Gene Name:
Tu translation elongation factor, mitochondrial
Gene Aliases:
COXPD4, EF-TuMT, EFTU, P43
Chromosome Location:
Chr.16: 28842411 - 28859562 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00944504_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TUFM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003321.4 NP_003312.3 1 - 2 195 110
XM_011545928.2 XP_011544230.1 1 - 2 289 110
XM_017023619.1 XP_016879108.1 2 - 3 452 110
GenBank mRNA BC001633.1 - 1 - 2 101 3536952 110
BC010041.2 - 1 - 2 109 3629164 110
S75463.1 - 1 - 2 147 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600009883.hg
This assay targets the same location as HTA transcript cluster(s) TC16000994.hg

Additional Information:

This assay is in the 5'UTR region of the BC001633.1,BC010041.2,NM_003321.4,S75463.1,XM_011545928.2,XM_017023619.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TUFM

Panther Classification:

Molecular Function -

translation elongation factor

Gene Ontology Categories:

Function(s) Process(es)

translational elongation
mitochondrial translational elongation
translation elongation factor activity
GTPase activity
protein binding
GTP binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700010137.hg.1 ENST00000581294.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16014312.hg.1 ENST00000313511.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16014312.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16014312.hg.1 TC16000994.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000561644.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000561644.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000561644.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000577590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000577590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NR_039872.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NR_039872.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16014311.hg.1 ENST00000313511.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16014311.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16014311.hg.1 TC16000994.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR16014314.hg.1 ENST00000313511.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16014314.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16014314.hg.1 TC16000994.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1600176062.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600176062.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600176062.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay PSR1600176062.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600176062.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600176062.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay PSR1600176062.hg.1 TC1600009883.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600176063.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600176063.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600176063.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay PSR1600176063.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600176063.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600176063.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay PSR1600176063.hg.1 TC1600009883.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000561644.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000561644.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000561644.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000577590.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000577590.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NR_039872.1 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NR_039872.1 Refseq
Affymetrix Human Clariom D Assay PSR1600176061.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600176061.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600176061.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay PSR1600176061.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600176061.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600176061.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay PSR1600176061.hg.1 TC1600009883.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600176066.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600176066.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay PSR1600176066.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600176066.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay PSR1600176066.hg.1 TC1600009883.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1700010137.hg.1 ENST00000581294.2 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0