This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00944508_g1
See other TUFM GE Assays ›
Gene Symbol:
TUFM
Gene Name:
Tu translation elongation factor, mitochondrial
Gene Aliases:
COXPD4, EF-TuMT, EFTU, P43
Chromosome Location:
Chr.16: 28842411 - 28859562 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00944508_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TUFM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003321.4 NP_003312.3 5 - 6 823 65
XM_011545928.2 XP_011544230.1 5 - 6 917 65
XM_017023619.1 XP_016879108.1 6 - 7 1080 65
GenBank mRNA BC001633.1 - 5 - 6 729 3536952 65
BC010041.2 - 5 - 6 737 3629164 65
L38995.1 - 5 - 6 682 65
S75463.1 - 5 - 6 766 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600009883.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000994.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TUFM

Panther Classification:

Molecular Function -

translation elongation factor

Gene Ontology Categories:

Function(s) Process(es)

translational elongation
mitochondrial translational elongation
translation elongation factor activity
GTPase activity
protein binding
GTP binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16014307.hg.1 ENST00000313511.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16014307.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16014307.hg.1 TC16000994.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000561644.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000561644.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000561644.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000577590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 ENST00000577590.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NR_039872.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009883.hg.1 NR_039872.1 Refseq
Affymetrix Human Clariom D Assay PSR1600176055.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600176055.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600176055.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay PSR1600176055.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600176055.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600176055.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay PSR1600176055.hg.1 TC1600009883.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000313511.7 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000561644.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000561644.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000561644.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000565012.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000569217.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000577590.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 ENST00000577590.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NM_003321.4 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NR_039872.1 Refseq
Affymetrix Human Clariom D Assay TC1600009883.hg.1 NR_039872.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0