This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00944963_g1
See other AGPAT2 GE Assays ›
Gene Symbol:
AGPAT2
Gene Name:
1-acylglycerol-3-phosphate O-acyltransferase 2
Gene Aliases:
1-AGPAT2, BSCL, BSCL1, LPAAB, LPAAT-beta
Chromosome Location:
Chr.9: 136673143 - 136687459 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00944963_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AGPAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006412.3 NP_006403.2 3 - 4 593 90
GenBank mRNA AF000237.1 - 3 - 4 557 90
AF011374.1 - 3 - 4 551 90
BC000026.2 - 3 - 4 536 3507150 90
BC007269.1 - 1 - 2 168 90
BC019292.1 - 3 - 4 545 2823580 90
BF347518.1 - 3 - 4 594 90
U56418.2 - 3 - 4 530 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900011992.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001738.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AGPAT2

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of cytokine production
positive regulation of cytokine-mediated signaling pathway
phospholipid metabolic process
phosphatidic acid biosynthetic process
epidermis development
CDP-diacylglycerol biosynthetic process
triglyceride biosynthetic process
response to antidepressant
1-acylglycerol-3-phosphate O-acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09022311.hg.1 TC09001738.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09022311.hg.1 ENST00000371696.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09022311.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09022311.hg.1 NM_006412.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000470861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000470861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000470861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000470861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000470861.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371694.7 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000470861.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000470861.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000470861.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000470861.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000470861.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_001012727.1 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay TC0900011992.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay PSR0900182230.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900182230.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900182230.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900182230.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay PSR0900182230.hg.1 ENST00000371696.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900182230.hg.1 ENST00000472820.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900182230.hg.1 ENST00000538402.1 Ensembl
Affymetrix Human Clariom D Assay PSR0900182230.hg.1 NM_006412.3 Refseq
Affymetrix Human Clariom D Assay PSR0900182230.hg.1 TC0900011992.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0