This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00945899_m1
See other DNMT1 GE Assays ›
Gene Symbol:
DNMT1
Gene Name:
DNA (cytosine-5-)-methyltransferase 1
Gene Aliases:
ADCADN, AIM, CXXC9, DNMT, HSN1E, MCMT, m.HsaI
Chromosome Location:
Chr.19: 10133344 - 10195135 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00945899_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DNMT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130823.2 NP_001124295.1 39 - 40 5008 98
NM_001318730.1 NP_001305659.1 38 - 39 4888 98
NM_001318731.1 NP_001305660.1 39 - 40 4887 98
NM_001379.3 NP_001370.1 38 - 39 4960 98
GenBank mRNA AB209413.2 - 38 - 39 4695 98
AF180682.1 - 36 - 37 4467 98
AK122759.1 - 11 - 12 2644 98
BC092517.1 - 39 - 40 4767 3077831 98
BC126227.1 - 39 - 40 4952 8991943 98
BC144093.1 - 39 - 40 4952 9052613 98
HY245803.1 - 2 - 3 98
X63692.1 - 38 - 39 4961 98

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900011860.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19001159.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DNMT1

Panther Classification:

Molecular Function -

DNA metabolism protein DNA methyltransferase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
DNA methylation
transcription, DNA-templated
Ras protein signal transduction
brain development
aging
response to heat
response to toxic substance
response to ionizing radiation
maintenance of DNA methylation
DNA methylation on cytosine within a CG sequence
positive regulation of gene expression
response to activity
gene silencing
covalent chromatin modification
neuron differentiation
response to caffeine
response to estradiol
response to lipopolysaccharide
response to vitamin A
response to testosterone
cellular response to platelet-derived growth factor stimulus
response to antidepressant
regulation of cell proliferation
DNA hypermethylation
response to ethanol
negative regulation of gene expression, epigenetic
S-adenosylhomocysteine metabolic process
S-adenosylmethioninamine metabolic process
S-adenosylmethionine metabolic process
positive regulation of histone H3-K4 methylation
negative regulation of histone H3-K9 methylation
cellular response to amino acid stimulus
cellular response to lead ion
cellular response to transforming growth factor beta stimulus
C-5 methylation of cytosine
positive regulation of methylation-dependent chromatin silencing
cellular response to nerve growth factor stimulus
DNA binding
double-stranded DNA binding
RNA binding
DNA (cytosine-5-)-methyltransferase activity
protein binding
zinc ion binding
methyl-CpG binding
DNA-methyltransferase activity
protein domain specific binding
estrogen receptor binding
histone deacetylase binding
unmethylated CpG binding
DNA (cytosine-5-)-methyltransferase activity, acting on CpG substrates
promoter-specific chromatin binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19016051.hg.1 ENST00000340748.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19016051.hg.1 ENST00000359526.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19016051.hg.1 ENST00000540357.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19016051.hg.1 ENST00000541266.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19016051.hg.1 NM_001130823.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19016051.hg.1 NM_001379.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19016051.hg.1 TC19001159.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 ENST00000340748.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 ENST00000359526.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 ENST00000540357.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 ENST00000586588.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 ENST00000588913.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 ENST00000591798.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 ENST00000592705.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 NM_001130823.2 Refseq
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 NM_001379.3 Refseq
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 TC1900011860.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 ENST00000340748.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 ENST00000359526.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 ENST00000540357.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 ENST00000586588.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 ENST00000588913.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 ENST00000591798.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 ENST00000592705.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 NM_001130823.2 Refseq
Affymetrix Human Clariom D Assay PSR1900203952.hg.1 NM_001379.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0