This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00946091_g1
See other EPX GE Assays ›
Gene Symbol:
EPX
Gene Name:
eosinophil peroxidase
Gene Aliases:
EPO, EPP, EPX-PEN, EPXD
Chromosome Location:
Chr.17: 58192724 - 58205174 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00946091_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EPX
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000502.5 NP_000493.1 2 - 3 297 106
GenBank mRNA BM147889.1 - 2 - 3 297 106
X14346.1 - 2 - 3 138 106

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700008424.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17000711.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EPX

Panther Classification:

Molecular Function -

peroxidase

Gene Ontology Categories:

Function(s) Process(es)

defense response to nematode
response to oxidative stress
negative regulation of interleukin-10 production
negative regulation of interleukin-5 production
positive regulation of interleukin-4 production
hydrogen peroxide catabolic process
oxidation-reduction process
eosinophil migration
cellular oxidant detoxification
peroxidase activity
heme binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR17009255.hg.1 ENST00000225371.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17009255.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17009255.hg.1 TC17000711.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR17009256.hg.1 ENST00000225371.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17009256.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17009256.hg.1 TC17000711.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17009254.hg.1 ENST00000225371.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17009254.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17009254.hg.1 TC17000711.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR17009257.hg.1 ENST00000225371.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17009257.hg.1 NM_000502.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17009257.hg.1 TC17000711.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1700162638.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700162638.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay PSR1700162638.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700162638.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay PSR1700162638.hg.1 TC1700008424.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700162640.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700162640.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay PSR1700162640.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700162640.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay PSR1700162640.hg.1 TC1700008424.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008424.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay TC1700008424.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay PSR1700162637.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700162637.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay PSR1700162637.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700162637.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay PSR1700162637.hg.1 TC1700008424.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700162639.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700162639.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay PSR1700162639.hg.1 ENST00000225371.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700162639.hg.1 NM_000502.5 Refseq
Affymetrix Human Clariom D Assay PSR1700162639.hg.1 TC1700008424.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0