This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00946416_m1
See other ECM2 GE Assays ›
Gene Symbol:
ECM2
Gene Name:
extracellular matrix protein 2
Gene Aliases:
-
Chromosome Location:
Chr.9: 92493547 - 92536092 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00946416_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ECM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001393.3 NP_001384.1 3 - 4 666 164
XM_017014376.1 XP_016869865.1 2 - 3 506 164
XM_017014377.1 XP_016869866.1 2 - 3 506 164
GenBank mRNA AB011792.1 - 3 - 4 552 164
AK312820.1 - 3 - 4 564 164
BC107493.1 - 2 - 3 509 40035036 164

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0900010800.hg.1
This assay targets the same location as HTA transcript cluster(s) TC09001337.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ECM2

Gene Ontology Categories:

Function(s) Process(es)

cell-matrix adhesion
axonogenesis
positive regulation of cell-substrate adhesion
extracellular matrix organization
integrin binding
heparin binding
collagen V binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR09016773.hg.1 TC09001337.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09016773.hg.1 ENST00000344604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09016773.hg.1 ENST00000444490.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09016773.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09016773.hg.1 NM_001197296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09016773.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR09016774.hg.1 TC09001337.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR09016774.hg.1 ENST00000344604.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR09016774.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000375540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000375540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000375540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000375540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000375540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000375540.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000395534.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000395534.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000395534.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000395534.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000395534.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000375540.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000375540.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000375540.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000375540.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000375540.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000375540.5 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000395534.2 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000395534.2 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000395534.2 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000395534.2 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000395534.2 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay TC0900010800.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay PSR0900173536.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900173536.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900173536.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay PSR0900173536.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay PSR0900173536.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay PSR0900173536.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900173536.hg.1 ENST00000444490.6 Ensembl
Affymetrix Human Clariom D Assay PSR0900173536.hg.1 NM_001197295.1 Refseq
Affymetrix Human Clariom D Assay PSR0900173536.hg.1 NM_001197296.1 Refseq
Affymetrix Human Clariom D Assay PSR0900173536.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay PSR0900173536.hg.1 TC0900010800.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0900173537.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900173537.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay PSR0900173537.hg.1 ENST00000344604.9 Ensembl
Affymetrix Human Clariom D Assay PSR0900173537.hg.1 NM_001393.3 Refseq
Affymetrix Human Clariom D Assay PSR0900173537.hg.1 TC0900010800.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0