This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00949505_g1
See other PCNP GE Assays ›
Gene Symbol:
PCNP
Gene Name:
PEST proteolytic signal containing nuclear protein
Gene Aliases:
-
Chromosome Location:
Chr.3: 101574094 - 101594465 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00949505_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCNP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320395.1 NP_001307324.1 2 - 3 404 90
NM_001320399.1 NP_001307328.1 3 - 4 530 90
NM_020357.2 NP_065090.1 2 - 3 404 90
NR_135223.1 2 - 3 352 90
NR_135227.1 2 - 3 605 90
NR_135228.1 2 - 3 352 90
NR_135232.1 3 - 4 447 90
GenBank mRNA AB037675.1 - 2 - 3 300 90
AK025551.1 - 2 - 3 244 90
AK314629.1 - 2 - 3 285 90
BC013916.1 - 2 - 3 312 3929908 90
BC022001.2 - 2 - 3 302 4823990 90
BC107796.1 - 2 - 3 246 90
BI547939.1 - 2 - 3 517 90
BI596913.1 - 3 - 4 455 90
CN482876.1 - 3 - 4 331 90
DA041324.1 - 2 - 3 260 90
DA304502.1 - 2 - 3 352 90

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PCNP

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
protein ubiquitination
proteasome-mediated ubiquitin-dependent protein catabolic process
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline