This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00950873_g1
See other TNFRSF18 GE Assays ›
Gene Symbol:
TNFRSF18
Gene Name:
TNF receptor superfamily member 18
Gene Aliases:
AITR, CD357, GITR, GITR-D
Chromosome Location:
Chr.1: 1203508 - 1207900 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00950873_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TNFRSF18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004195.2 NP_004186.1 3 - 4 539 63
NM_148902.1 NP_683700.1 3 - 4 539 63
XM_017002722.1 XP_016858211.1 3 - 4 1730 63
GenBank mRNA AF117297.1 - 3 - 4 401 63
AF125304.1 - 3 - 4 401 63
AY358877.1 - 3 - 4 521 63
BC152381.1 - 3 - 4 404 100013440 63
BC152386.1 - 3 - 4 404 63
BT019531.1 - 3 - 4 401 63
BT019532.1 - 3 - 4 401 63
KP067911.1 - 3 - 4 539 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100012452.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01002085.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TNFRSF18

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of leukocyte migration
inflammatory response
immune response
signal transduction
multicellular organism development
extrinsic apoptotic signaling pathway via death domain receptors
response to lipopolysaccharide
tumor necrosis factor-mediated signaling pathway
regulation of cell proliferation
positive regulation of tyrosine phosphorylation of Stat1 protein
negative regulation of apoptotic process
positive regulation of cell adhesion
tumor necrosis factor-activated receptor activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01031537.hg.1 TC01002085.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01031537.hg.1 ENST00000379265.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01031537.hg.1 ENST00000379268.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01031537.hg.1 ENST00000486728.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01031537.hg.1 NM_004195.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01031537.hg.1 NM_148902.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000328596.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000328596.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000328596.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000328596.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000328596.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000328596.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000379265.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000379265.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000379265.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000379265.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000379265.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000486728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000486728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000486728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000486728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 ENST00000486728.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_148901.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_148901.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_148901.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_148901.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_148901.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_148901.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_148901.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000328596.10 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000328596.10 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000328596.10 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000328596.10 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000328596.10 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000328596.10 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000379265.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000379265.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000379265.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000379265.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000379265.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000486728.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000486728.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000486728.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000486728.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 ENST00000486728.5 Ensembl
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_004195.2 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_148901.1 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_148901.1 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_148901.1 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_148901.1 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_148901.1 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_148901.1 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_148901.1 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Clariom D Assay TC0100012452.hg.1 NM_148902.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191473.hg.1 ENST00000379265.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191473.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191473.hg.1 ENST00000486728.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191473.hg.1 NM_004195.2 Refseq
Affymetrix Human Clariom D Assay PSR0100191473.hg.1 NM_148902.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191473.hg.1 ENST00000379265.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191473.hg.1 ENST00000379268.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100191473.hg.1 ENST00000486728.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100191473.hg.1 NM_004195.2 Refseq
Affymetrix Human Clariom D Assay PSR0100191473.hg.1 NM_148902.1 Refseq
Affymetrix Human Clariom D Assay PSR0100191473.hg.1 TC0100012452.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0