This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00953555_g1
See other UQCRH GE Assays ›
Gene Symbol:
UQCRH
Gene Name:
ubiquinol-cytochrome c reductase hinge protein
Gene Aliases:
QCR6, UQCR8
Chromosome Location:
Chr.1: 46303613 - 46316777 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00953555_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UQCRH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001297565.1 NP_001284494.1 3 - 4 387 88
GenBank mRNA BC093060.1 - 3 - 4 302 30409772 88
BP366403.1 - 3 - 4 389 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

oxidative phosphorylation
mitochondrial electron transport, ubiquinol to cytochrome c
aerobic respiration
oxidation-reduction process
hydrogen ion transmembrane transport
protein binding
ubiquinol-cytochrome-c reductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline