This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00953719_g1
See other FABP7 GE Assays ›
Gene Symbol:
FABP7
Gene Name:
fatty acid binding protein 7
Gene Aliases:
B-FABP, BLBP, FABPB, MRG
Chromosome Location:
Chr.6: 122749201 - 122784077 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00953719_g1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FABP7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001319039.1 NP_001305968.1 2 - 3 568 122
NM_001319042.1 NP_001305971.1 2 - 3 309 122
NM_001446.4 NP_001437.1 2 - 3 568 122
GenBank mRNA AJ002962.1 - 2 - 3 324 122
AK289836.1 - 2 - 3 327 122
AK311867.1 - 2 - 3 327 122
AL512688.1 - 2 - 3 284 122
AW156936.1 - 2 - 3 122
BC012299.1 - 2 - 3 434 4707233 122
BU736791.1 - 2 - 3 122
CR457057.1 - 2 - 3 248 122
D50373.1 - 2 - 3 300 122
D88648.1 - 2 - 3 299 122
KM111186.1 - 2 - 3 309 122
U51338.1 - 2 - 3 303 122
U81235.1 - 2 - 3 263 122

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FABP7

Gene Ontology Categories:

Function(s) Process(es)

transport
nervous system development
negative regulation of cell proliferation
triglyceride catabolic process
cell proliferation in forebrain
neurogenesis
epithelial cell proliferation
prepulse inhibition
transporter activity
lipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 ENST00000356535.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 ENST00000356535.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 ENST00000356535.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 ENST00000356535.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 ENST00000356535.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 NM_001446.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 NM_001446.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 NM_001446.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 NM_001446.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 NM_001446.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 NM_001446.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 NM_001446.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600009332.hg.1 NM_001446.4 Refseq
Affymetrix Human Clariom D Assay TC0600009332.hg.1 ENST00000356535.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009332.hg.1 ENST00000356535.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009332.hg.1 ENST00000356535.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009332.hg.1 ENST00000356535.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009332.hg.1 ENST00000356535.4 Ensembl
Affymetrix Human Clariom D Assay TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Clariom D Assay TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Clariom D Assay TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Clariom D Assay TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Clariom D Assay TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Clariom D Assay TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Clariom D Assay TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Clariom D Assay TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Clariom D Assay TC0600009332.hg.1 ENST00000368444.7 Ensembl
Affymetrix Human Clariom D Assay TC0600009332.hg.1 NM_001446.4 Refseq
Affymetrix Human Clariom D Assay TC0600009332.hg.1 NM_001446.4 Refseq
Affymetrix Human Clariom D Assay TC0600009332.hg.1 NM_001446.4 Refseq
Affymetrix Human Clariom D Assay TC0600009332.hg.1 NM_001446.4 Refseq
Affymetrix Human Clariom D Assay TC0600009332.hg.1 NM_001446.4 Refseq
Affymetrix Human Clariom D Assay TC0600009332.hg.1 NM_001446.4 Refseq
Affymetrix Human Clariom D Assay TC0600009332.hg.1 NM_001446.4 Refseq
Affymetrix Human Clariom D Assay TC0600009332.hg.1 NM_001446.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0