This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00955708_g1
See other GLTP GE Assays ›
Gene Symbol:
GLTP
Gene Name:
glycolipid transfer protein
Gene Aliases:
-
Chromosome Location:
Chr.12: 109850943 - 109880488 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00955708_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLTP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016433.3 NP_057517.1 1 - 2 215 87
XM_011538434.2 XP_011536736.1 1 - 2 168 87
GenBank mRNA AF209704.1 - 1 - 2 111 87
AK313457.1 - 1 - 2 104 87
AY372530.1 - 1 - 2 101 87
AY372531.1 - 1 - 2 101 87
AY372532.1 - 1 - 2 130 87
BC009932.2 - 1 - 2 215 3959556 87
JN039379.1 - 1 - 2 101 87

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200011885.hg
This assay targets the same location as HTA transcript cluster(s) TC12001958.hg

Additional Information:

This assay is in the 5'UTR region of the XM_011538434.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GLTP

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

glycosphingolipid metabolic process
glycolipid transport
protein binding
lipid binding
glycolipid transporter activity
glycolipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100006990.hg.1 ENST00000528859.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12025448.hg.1 ENST00000318348.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12025448.hg.1 ENST00000544393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12025448.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12025448.hg.1 TC12001958.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12025449.hg.1 ENST00000318348.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12025449.hg.1 ENST00000544393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12025449.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12025449.hg.1 TC12001958.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR12025450.hg.1 ENST00000318348.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12025450.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12025450.hg.1 TC12001958.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay PSR1200192216.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200192216.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192216.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay PSR1200192216.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200192216.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192216.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay PSR1200192216.hg.1 TC1200011885.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200192217.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200192217.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay PSR1200192217.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200192217.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay PSR1200192217.hg.1 TC1200011885.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200192213.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200192213.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192213.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200192213.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192213.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200192213.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay PSR1200192213.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200192213.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192213.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200192213.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192213.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200192213.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay PSR1200192213.hg.1 TC1200011885.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100006990.hg.1 ENST00000528859.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200192211.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200192211.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192211.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200192211.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200192211.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192211.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200192211.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay PSR1200192211.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200192211.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192211.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200192211.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200192211.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192211.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200192211.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay PSR1200192211.hg.1 TC1200011885.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200192212.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200192212.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192212.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200192212.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200192212.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192212.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200192212.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay PSR1200192212.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200192212.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192212.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200192212.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200192212.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192212.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200192212.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay PSR1200192212.hg.1 TC1200011885.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline