This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00955709_g1
See other GLTP GE Assays ›
Gene Symbol:
GLTP
Gene Name:
glycolipid transfer protein
Gene Aliases:
-
Chromosome Location:
Chr.12: 109850943 - 109880488 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00955709_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GLTP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016433.3 NP_057517.1 2 - 3 278 66
GenBank mRNA AF209704.1 - 2 - 3 174 66
AK313457.1 - 2 - 3 167 66
AY372530.1 - 2 - 3 164 66
AY372531.1 - 2 - 3 164 66
AY372532.1 - 2 - 3 193 66
BC009932.2 - 2 - 3 278 3959556 66

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200011885.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12001958.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GLTP

Panther Classification:

Molecular Function -

transfer/carrier protein

Gene Ontology Categories:

Function(s) Process(es)

glycosphingolipid metabolic process
glycolipid transport
protein binding
lipid binding
glycolipid transporter activity
glycolipid binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12025447.hg.1 ENST00000318348.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12025447.hg.1 ENST00000544393.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12025447.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12025447.hg.1 TC12001958.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay TC1200011885.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay PSR1200192208.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200192208.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192208.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200192208.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200192208.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192208.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200192208.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay PSR1200192208.hg.1 ENST00000318348.8 Ensembl
Affymetrix Human Clariom D Assay PSR1200192208.hg.1 ENST00000536390.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192208.hg.1 ENST00000537066.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200192208.hg.1 ENST00000540772.1 Ensembl
Affymetrix Human Clariom D Assay PSR1200192208.hg.1 ENST00000544393.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200192208.hg.1 ENST00000617457.4 Ensembl
Affymetrix Human Clariom D Assay PSR1200192208.hg.1 NM_016433.3 Refseq
Affymetrix Human Clariom D Assay PSR1200192208.hg.1 TC1200011885.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-kg5vd:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0