Assay Details
| Gene Symbol: GLTP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016433.3 | NP_057517.1 | 3 - 4 | 415 | 62 | |
| XM_011538434.2 | XP_011536736.1 | 4 - 5 | 707 | 62 | ||
| GenBank mRNA | AF209704.1 | - | 3 - 4 | 311 | 62 | |
| AK313457.1 | - | 3 - 4 | 304 | 62 | ||
| AY372530.1 | - | 3 - 4 | 301 | 62 | ||
| AY372531.1 | - | 3 - 4 | 301 | 62 | ||
| AY372532.1 | - | 3 - 4 | 330 | 62 | ||
| BC009932.2 | - | 3 - 4 | 415 | 3959556 | 62 | |
| JN039379.1 | - | 4 - 5 | 640 | 62 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001958.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12025445.hg.1 | ENST00000318348.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025445.hg.1 | ENST00000544393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025445.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025445.hg.1 | TC12001958.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025446.hg.1 | ENST00000318348.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025446.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12025446.hg.1 | TC12001958.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200192205.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192205.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192205.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192205.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192205.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192205.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192205.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200192205.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192205.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192205.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192205.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192205.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192205.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192205.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200192205.hg.1 | TC1200011885.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200192207.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192207.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192207.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192207.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192207.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192207.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200192207.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192207.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192207.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192207.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192207.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192207.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200192207.hg.1 | TC1200011885.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000540772.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200011885.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200192206.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192206.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192206.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192206.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192206.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192206.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200192206.hg.1 | ENST00000318348.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192206.hg.1 | ENST00000536390.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192206.hg.1 | ENST00000537066.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192206.hg.1 | ENST00000544393.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192206.hg.1 | ENST00000617457.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200192206.hg.1 | NM_016433.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200192206.hg.1 | TC1200011885.hg.1 | Clariom D TC |

GLTP
Genomic Map