This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00956619_m1
See other ELAVL4 GE Assays ›
Gene Symbol:
ELAVL4
Gene Name:
ELAV like neuron-specific RNA binding protein 4
Gene Aliases:
HUD, PNEM
Chromosome Location:
Chr.1: 50048014 - 50203786 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00956619_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ELAVL4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001324209.1 NP_001311138.1 2 - 3 373 122
NM_001324212.1 NP_001311141.1 2 - 3 373 122
NM_001324214.1 NP_001311143.1 2 - 3 443 122
NR_136725.1 2 - 3 443 122
XM_011540894.2 XP_011539196.1 2 - 3 440 122
XM_011540895.2 XP_011539197.1 2 - 3 344 122
GenBank mRNA CK005107.1 - 2 - 3 376 122

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001324209.1, NM_001324212.1, NM_001324214.1, XM_011540894.2, XM_011540895.2 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Inventoried Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

RNA processing
mRNA processing
nucleotide binding
RNA binding
mRNA 3'-UTR binding
AU-rich element binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tks2p:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0