Assay Details
| Gene Symbol: ZMPSTE24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005857.4 | NP_005848.2 | 1 - 2 | 346 | 73 | |
| GenBank mRNA | AB016068.1 | - | 1 - 2 | 302 | 73 | |
| AF064867.1 | - | 1 - 2 | 247 | 73 | ||
| AK074784.1 | - | 1 - 2 | 202 | 73 | ||
| AK075007.1 | - | 1 - 2 | 225 | 73 | ||
| BC037283.1 | - | 1 - 2 | 250 | 5269064 | 73 | |
| DA398037.1 | - | 1 - 2 | 318 | 73 | ||
| DB096663.1 | - | 1 - 2 | 348 | 73 | ||
| DB285015.1 | - | 1 - 2 | 335 | 73 | ||
| Y13834.1 | - | 1 - 2 | 157 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000508.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01007923.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007923.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007923.hg.1 | TC01000508.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007927.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007927.hg.1 | ENST00000479131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007927.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007927.hg.1 | TC01000508.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000474142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000474142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000474142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007921.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007921.hg.1 | TC01000508.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007922.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007922.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007922.hg.1 | TC01000508.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007925.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007925.hg.1 | ENST00000479131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007925.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01007925.hg.1 | TC01000508.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0100155945.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155945.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100155945.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155945.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100155945.hg.1 | TC0100007949.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100155950.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155950.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155950.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155950.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100155950.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155950.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155950.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155950.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100155950.hg.1 | TC0100007949.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100155947.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155947.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155947.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155947.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100155947.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155947.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155947.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155947.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100155947.hg.1 | TC0100007949.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100155948.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155948.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155948.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155948.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100155948.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155948.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155948.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155948.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100155948.hg.1 | TC0100007949.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000472583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000474142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000474142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000474142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | ENST00000479131.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100007949.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100155944.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155944.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100155944.hg.1 | ENST00000372759.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100155944.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100155944.hg.1 | TC0100007949.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100155943.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100155943.hg.1 | NM_005857.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100155943.hg.1 | TC0100007949.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 146073 | - | - |
| ABI Human Genome Survey Microarray Version 2.0 | 180316 | - | - |

ZMPSTE24
Genomic Map