| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000472583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000472583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000472583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000472583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000472583.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000474142.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000474142.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000474142.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000479131.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000479131.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000479131.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000479131.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
ENST00000479131.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007929.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007929.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007929.hg.1 |
TC01000508.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007930.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007930.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01007930.hg.1 |
TC01000508.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000472583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000472583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000472583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000472583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000472583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000474142.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000474142.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000474142.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000479131.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000479131.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000479131.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000479131.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
ENST00000479131.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100007949.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100155952.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155952.hg.1 |
ENST00000472583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155952.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100155952.hg.1 |
ENST00000372759.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155952.hg.1 |
ENST00000472583.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100155952.hg.1 |
NM_005857.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100155952.hg.1 |
TC0100007949.hg.1 |
Clariom D TC |