This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00956774_m1
See other ZMPSTE24 GE Assays ›
Gene Symbol:
ZMPSTE24
Gene Name:
zinc metallopeptidase STE24
Gene Aliases:
FACE-1, FACE1, HGPS, PRO1, STE24, Ste24p
Chromosome Location:
Chr.1: 40258050 - 40294184 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00956774_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZMPSTE24
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005857.4 NP_005848.2 5 - 6 845 156
GenBank mRNA AB016068.1 - 5 - 6 801 156
AF064867.1 - 5 - 6 746 156
AK074784.1 - 5 - 6 701 156
AK075007.1 - 5 - 6 724 156
BC037283.1 - 5 - 6 749 5269064 156
Y13834.1 - 5 - 6 656 156

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007949.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000508.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZMPSTE24

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
nuclear envelope organization
prenylated protein catabolic process
CAAX-box protein processing
metalloendopeptidase activity
metalloexopeptidase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01007934.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007934.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01007934.hg.1 TC01000508.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01007933.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01007933.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01007933.hg.1 TC01000508.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000472583.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000472583.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000472583.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000472583.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000472583.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000474142.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000474142.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000474142.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000479131.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000479131.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000479131.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000479131.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 ENST00000479131.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay PSR0100155956.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100155956.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay PSR0100155956.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100155956.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay PSR0100155956.hg.1 TC0100007949.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100155957.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100155957.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay PSR0100155957.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100155957.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay PSR0100155957.hg.1 TC0100007949.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000372759.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000472583.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000472583.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000472583.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000472583.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000472583.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000474142.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000474142.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000474142.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000479131.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000479131.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000479131.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000479131.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 ENST00000479131.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq
Affymetrix Human Clariom D Assay TC0100007949.hg.1 NM_005857.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline