This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00956789_m1
See other GTF2H3 GE Assays ›
Gene Symbol:
GTF2H3
Gene Name:
general transcription factor IIH subunit 3
Gene Aliases:
BTF2, P34, TFB4, TFIIH
Chromosome Location:
Chr.12: 123633739 - 123662604 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00956789_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF2H3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001271867.1 NP_001258796.1 4 - 5 464 117
NM_001271868.1 NP_001258797.1 4 - 5 380 117
NM_001516.4 NP_001507.2 5 - 6 544 117
GenBank mRNA AK298128.1 - 4 - 5 374 117
AK313200.1 - 5 - 6 454 117
AL558542.3 - 5 - 6 501 117
BC031030.1 - 2 - 3 205 117
BC039726.2 - 4 - 5 410 117
BC047868.2 - 4 - 5 410 117
BC065250.1 - 5 - 6 437 6141067 117
DB482432.1 - 4 - 5 394 117
Z30093.1 - 4 - 5 423 117

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001271868.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GTF2H3

Panther Classification:

Molecular Function -

RNA metabolism protein nucleic acid metabolism protein general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
transcription-coupled nucleotide-excision repair
nucleotide-excision repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
transcription initiation from RNA polymerase I promoter
transcription elongation from RNA polymerase I promoter
termination of RNA polymerase I transcription
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
7-methylguanosine mRNA capping
translation
nucleotide-excision repair, DNA incision
phosphorylation of RNA polymerase II C-terminal domain
global genome nucleotide-excision repair
damaged DNA binding
transcription factor activity, sequence-specific DNA binding
protein kinase activity
protein binding
DNA-dependent ATPase activity
translation factor activity, RNA binding
RNA polymerase II carboxy-terminal domain kinase activity
metal ion binding
protein N-terminus binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline