This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00957795_m1
See other PLAGL1 GE Assays ›
Gene Symbol:
PLAGL1
Gene Name:
PLAG1 like zinc finger 1
Gene Aliases:
LOT1, ZAC, ZAC1
Chromosome Location:
Chr.6: 143940300 - 144064599 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00957795_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PLAGL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080955.2 NP_001074424.1 5 - 6 360 78
NM_001080956.2 NP_001074425.1 5 - 6 555 78
NM_001289037.1 NP_001275966.1 4 - 5 319 78
NM_001289038.1 NP_001275967.1 3 - 4 273 78
NM_001289040.1 NP_001275969.1 5 - 6 345 78
NM_001289041.1 NP_001275970.1 5 - 6 555 78
NM_001317158.1 NP_001304087.1 2 - 3 775 78
NM_001317160.1 NP_001304089.1 5 - 6 541 78
GenBank mRNA AJ303119.1 - 1 - 2 83 78
AK314570.1 - 5 - 6 402 78
BX537397.1 - 2 - 3 3744 78
DA426770.1 - 5 - 6 402 78
DA549753.1 - 5 - 6 506 78
DA675185.1 - 5 - 6 354 78
DA717476.1 - 5 - 6 395 78
U81992.1 - 2 - 3 167 78

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AJ303119.1, AK314570.1, BX537397.1, NM_001080955.2, NM_001080956.2, NM_001289037.1, NM_001289038.1, NM_001289040.1, NM_001289041.1, NM_001317158.1, NM_001317160.1, U81992.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription from RNA polymerase II promoter
apoptotic process
DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
cell cycle arrest
cell differentiation
skeletal muscle cell differentiation
positive regulation of transcription from RNA polymerase II promoter
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0