This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00958209_g1
See other APBB1IP GE Assays ›
Gene Symbol:
APBB1IP
Gene Name:
amyloid beta precursor protein binding family B member 1 interacting protein
Gene Aliases:
INAG1, PREL1, RARP1, RIAM
Chromosome Location:
Chr.10: 26438324 - 26567879 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00958209_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APBB1IP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019043.3 NP_061916.3 1 - 2 123 103
XM_011519514.2 XP_011517816.1 1 - 2 136 103
GenBank mRNA AK303597.1 - 1 - 2 683 103
AK308916.1 - 1 - 2 104 103
BM150270.1 - 1 - 2 124 103
BM556115.1 - 90 103

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000007072.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10000181.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK303597.1,NM_019043.3,XM_011519514.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene APBB1IP

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
signal transduction
positive regulation of cell adhesion
protein binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10002155.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002155.hg.1 ENST00000376236.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002155.hg.1 ENST00000445780.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002155.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10002155.hg.1 TC10000181.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000362863.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000493857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000493857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 ENST00000493857.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10002156.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002156.hg.1 ENST00000376236.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002156.hg.1 ENST00000445780.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002156.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10002156.hg.1 TC10000181.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10002153.hg.1 ENST00000376236.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002153.hg.1 ENST00000445780.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002153.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10002153.hg.1 TC10000181.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10002157.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002157.hg.1 ENST00000376236.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002157.hg.1 ENST00000445780.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002157.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10002157.hg.1 TC10000181.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10002154.hg.1 ENST00000376236.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002154.hg.1 ENST00000445780.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10002154.hg.1 NM_019043.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10002154.hg.1 TC10000181.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000148234.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000148234.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000148234.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay PSR1000148234.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000148234.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000148234.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay PSR1000148234.hg.1 TC1000007072.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000362863.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000493857.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000493857.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 ENST00000493857.1 Ensembl
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay TC1000007072.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay PSR1000148231.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000148231.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay PSR1000148231.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000148231.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay PSR1000148231.hg.1 TC1000007072.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000148232.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000148232.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000148232.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay PSR1000148232.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000148232.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000148232.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay PSR1000148232.hg.1 TC1000007072.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000148233.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000148233.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000148233.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay PSR1000148233.hg.1 ENST00000356785.4 Ensembl
Affymetrix Human Clariom D Assay PSR1000148233.hg.1 ENST00000376236.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000148233.hg.1 NM_019043.3 Refseq
Affymetrix Human Clariom D Assay PSR1000148233.hg.1 TC1000007072.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 132885 - -
ABI Human Genome Survey Microarray Version 2.0 202785 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0