This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00958421_g1
See other CHD4 GE Assays ›
Gene Symbol:
CHD4
Gene Name:
chromodomain helicase DNA binding protein 4
Gene Aliases:
CHD-4, Mi-2b, Mi2-BETA
Chromosome Location:
Chr.12: 6570082 - 6607439 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00958421_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHD4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001273.3 NP_001264.2 30 - 31 4727 81
NM_001297553.1 NP_001284482.1 29 - 30 4572 81
XM_005253668.3 XP_005253725.1 30 - 31 4658 81
XM_006718958.1 XP_006719021.1 29 - 30 4817 81
XM_006718959.1 XP_006719022.1 30 - 31 4733 81
XM_006718960.1 XP_006719023.1 30 - 31 4730 81
XM_006718961.2 XP_006719024.1 30 - 31 4658 81
XM_006718962.1 XP_006719025.1 30 - 31 4694 81
XM_017018725.1 XP_016874214.1 30 - 31 4733 81
XM_017018726.1 XP_016874215.1 30 - 31 4733 81
XM_017018727.1 XP_016874216.1 30 - 31 4730 81
XM_017018728.1 XP_016874217.1 30 - 31 4730 81
XM_017018729.1 XP_016874218.1 30 - 31 4658 81
XM_017018730.1 XP_016874219.1 30 - 31 4694 81
XM_017018731.1 XP_016874220.1 30 - 31 4694 81
XM_017018732.1 XP_016874221.1 30 - 31 4619 81
XM_017018733.1 XP_016874222.1 30 - 31 4619 81
XM_017018734.1 XP_016874223.1 30 - 31 4619 81
GenBank mRNA AK128741.1 - 29 - 30 4617 81
BC038596.1 - 29 - 30 4752 5528023 81
CR936680.1 - 29 - 30 4480 81
X86691.1 - 29 - 30 4604 81

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200009742.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12001143.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CHD4

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
histone deacetylation
DNA duplex unwinding
ATP-dependent chromatin remodeling
spindle assembly
terminal button organization
regulation of signal transduction by p53 class mediator
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II repressing transcription factor binding
DNA binding
ATP-dependent DNA helicase activity
histone deacetylase activity
protein binding
ATP binding
microtubule binding
zinc ion binding
nucleosomal DNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12015034.hg.1 ENST00000309577.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12015034.hg.1 ENST00000357008.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12015034.hg.1 ENST00000537464.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12015034.hg.1 ENST00000544040.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12015034.hg.1 ENST00000544484.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12015034.hg.1 NM_001273.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12015034.hg.1 TC12001143.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1200172868.hg.1 ENST00000357008.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200172868.hg.1 ENST00000544040.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200172868.hg.1 ENST00000544484.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200172868.hg.1 NM_001273.3 Refseq
Affymetrix Human Clariom D Assay PSR1200172868.hg.1 NM_001297553.1 Refseq
Affymetrix Human Clariom D Assay PSR1200172868.hg.1 ENST00000357008.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200172868.hg.1 ENST00000544040.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200172868.hg.1 ENST00000544484.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200172868.hg.1 NM_001273.3 Refseq
Affymetrix Human Clariom D Assay PSR1200172868.hg.1 NM_001297553.1 Refseq
Affymetrix Human Clariom D Assay PSR1200172868.hg.1 TC1200009742.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1200172869.hg.1 ENST00000357008.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200172869.hg.1 ENST00000544040.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200172869.hg.1 ENST00000544484.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200172869.hg.1 NM_001273.3 Refseq
Affymetrix Human Clariom D Assay PSR1200172869.hg.1 NM_001297553.1 Refseq
Affymetrix Human Clariom D Assay PSR1200172869.hg.1 ENST00000357008.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200172869.hg.1 ENST00000544040.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200172869.hg.1 ENST00000544484.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200172869.hg.1 NM_001273.3 Refseq
Affymetrix Human Clariom D Assay PSR1200172869.hg.1 NM_001297553.1 Refseq
Affymetrix Human Clariom D Assay PSR1200172869.hg.1 TC1200009742.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0