This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00960247_g1
See other POLR2G GE Assays ›
Gene Symbol:
POLR2G
Gene Name:
RNA polymerase II subunit G
Gene Aliases:
RPB19, RPB7, hRPB19, hsRPB7
Chromosome Location:
Chr.11: 62761539 - 62766715 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00960247_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR2G
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002696.2 NP_002687.1 6 - 7 583 63
GenBank mRNA AK312131.1 - 6 - 7 542 63
AW016747.1 - 2 - 3 63
BC112162.1 - 6 - 7 522 8327630 63
BC112164.1 - 6 - 7 522 8327632 63
U20659.1 - 6 - 7 501 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100007874.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11000563.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POLR2G

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, exonucleolytic
mRNA splicing, via spliceosome
transcription-coupled nucleotide-excision repair
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
7-methylguanosine mRNA capping
apoptotic process
fibroblast growth factor receptor signaling pathway
gene expression
gene silencing by RNA
somatic stem cell population maintenance
snRNA transcription from RNA polymerase II promoter
positive regulation of translational initiation
positive regulation of viral transcription
positive regulation of nuclear-transcribed mRNA poly(A) tail shortening
single-stranded DNA binding
single-stranded RNA binding
DNA-directed RNA polymerase activity
translation initiation factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11006796.hg.1 ENST00000301788.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11006796.hg.1 NM_002696.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11006796.hg.1 TC11000563.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000531996.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000533442.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000533442.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 ENST00000533442.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 NM_002696.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 NM_002696.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 NM_002696.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 NM_002696.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 NM_002696.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 NM_002696.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007874.hg.1 NM_002696.2 Refseq
Affymetrix Human Clariom D Assay PSR1100156076.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Clariom D Assay PSR1100156076.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100156076.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100156076.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100156076.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100156076.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100156076.hg.1 NM_002696.2 Refseq
Affymetrix Human Clariom D Assay PSR1100156076.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Clariom D Assay PSR1100156076.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100156076.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100156076.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100156076.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100156076.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100156076.hg.1 NM_002696.2 Refseq
Affymetrix Human Clariom D Assay PSR1100156076.hg.1 TC1100007874.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000301788.11 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000524819.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000525455.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000526368.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000527435.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531944.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531996.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531996.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531996.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531996.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531996.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000531996.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000533442.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000533442.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 ENST00000533442.5 Ensembl
Affymetrix Human Clariom D Assay TC1100007874.hg.1 NM_002696.2 Refseq
Affymetrix Human Clariom D Assay TC1100007874.hg.1 NM_002696.2 Refseq
Affymetrix Human Clariom D Assay TC1100007874.hg.1 NM_002696.2 Refseq
Affymetrix Human Clariom D Assay TC1100007874.hg.1 NM_002696.2 Refseq
Affymetrix Human Clariom D Assay TC1100007874.hg.1 NM_002696.2 Refseq
Affymetrix Human Clariom D Assay TC1100007874.hg.1 NM_002696.2 Refseq
Affymetrix Human Clariom D Assay TC1100007874.hg.1 NM_002696.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hwq2n:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0