This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00960512_m1
See other ABCC2 GE Assays ›
Gene Symbol:
ABCC2
Gene Name:
ATP binding cassette subfamily C member 2
Gene Aliases:
ABC30, CMOAT, DJS, MRP2, cMRP
Chromosome Location:
Chr.10: 99782598 - 99853741 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00960512_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABCC2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000392.4 NP_000383.1 6 - 7 876 69
XM_006717631.3 XP_006717694.1 6 - 7 853 69
XM_011539291.2 XP_011537593.1 6 - 7 852 69
XM_017015675.1 XP_016871164.1 6 - 7 853 69
GenBank mRNA BC136419.1 - 6 - 7 664 9020407 69
U49248.1 - 6 - 7 730 69
U63970.1 - 6 - 7 666 69
U66683.1 - 6 - 7 732 69
X96395.2 - 6 - 7 666 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000008628.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ABCC2

Panther Classification:

Molecular Function -

ATP-binding cassette (ABC) transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
drug transmembrane transport
response to oxidative stress
response to heat
bilirubin transport
prostaglandin transport
cellular chloride ion homeostasis
response to methotrexate
response to estrogen
response to arsenic-containing substance
response to steroid hormone
transmembrane transport
thyroid hormone transport
anion transmembrane transport
protein binding
ATP binding
organic anion transmembrane transporter activity
bilirubin transmembrane transporter activity
protein domain specific binding
ATPase activity, coupled to transmembrane movement of substances
anion transmembrane-transporting ATPase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000496621.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000496621.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000496621.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000496621.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 ENST00000496621.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay PSR1000158843.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000158843.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000158843.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay PSR1000158843.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000158843.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay PSR1000158843.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay PSR1000158843.hg.1 TC1000008628.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370434.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000370449.8 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000496621.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000496621.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000496621.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000496621.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 ENST00000496621.1 Ensembl
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq
Affymetrix Human Clariom D Assay TC1000008628.hg.1 NM_000392.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mmql6:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0