This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00960783_m1
See other PRSS33 GE Assays ›
Gene Symbol:
PRSS33
Gene Name:
protease, serine 33
Gene Aliases:
EOS
Chromosome Location:
Chr.16: 2783953 - 2787759 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00960783_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS33
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BC036846.1 - 1 - 2 80 5245847 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600009138.hg.1

Additional Information:

This assay is in the 5'UTR region of the BC036846.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

serine protease

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay PSR1600169606.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay PSR1600169606.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169606.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169606.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169606.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600169606.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay PSR1600169606.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay PSR1600169606.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169606.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169606.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169606.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600169606.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay PSR1600169606.hg.1 TC1600009138.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay PSR1600169611.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169611.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169611.hg.1 TC1600009138.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0