This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00960784_g1
See other PRSS33 GE Assays ›
Gene Symbol:
PRSS33
Gene Name:
protease, serine 33
Gene Aliases:
EOS
Chromosome Location:
Chr.16: 2783953 - 2787759 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00960784_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS33
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_152891.2 NP_690851.2 2 - 3 237 65
XM_011522451.2 XP_011520753.1 3 - 4 361 65
XM_011522452.2 XP_011520754.1 3 - 4 173 65
GenBank mRNA AF536382.1 - 2 - 3 145 65
BC036846.1 - 3 - 4 219 5245847 65
BC062334.1 - 2 - 3 239 6110198 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600009138.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000792.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PRSS33

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

proteolysis
serine-type endopeptidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16011612.hg.1 ENST00000293851.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16011612.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16011612.hg.1 TC16000792.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay PSR1600169603.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay PSR1600169603.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169603.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169603.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169603.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600169603.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay PSR1600169603.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay PSR1600169603.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169603.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600169603.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600169603.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600169603.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay PSR1600169603.hg.1 TC1600009138.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000293851.9 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000570702.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000571674.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000576886.5 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 ENST00000612593.4 Ensembl
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq
Affymetrix Human Clariom D Assay TC1600009138.hg.1 NM_152891.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0