This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00961742_m1
See other MYDGF GE Assays ›
Gene Symbol:
MYDGF
Gene Name:
myeloid derived growth factor
Gene Aliases:
C19orf10, EUROIMAGE1875335, IL25, IL27, IL27w, R33729_1, SF20
Chromosome Location:
Chr.19: 4657545 - 4670403 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00961742_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MYDGF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019107.3 NP_061980.1 4 - 5 437 140
GenBank mRNA AF282264.1 - 4 - 5 368 140
AK098829.1 - 4 - 5 376 140
AL365373.1 - 4 - 5 359 140
AL365374.1 - 4 - 5 140
BC003639.2 - 4 - 5 355 140
BC010129.2 - 4 - 5 404 3953027 140
BC014655.1 - 4 - 5 375 4562455 140

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900009357.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19001067.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MYDGF

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
positive regulation of protein phosphorylation
positive regulation of endothelial cell proliferation
apoptotic process
positive regulation of phosphatidylinositol 3-kinase signaling
IRE1-mediated unfolded protein response
negative regulation of apoptotic process
positive regulation of MAPK cascade
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein kinase B signaling
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19014598.hg.1 ENST00000262947.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19014598.hg.1 NM_019107.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19014598.hg.1 TC19001067.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000596031.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay TC1900009357.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay PSR1900174771.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900174771.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900174771.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900174771.hg.1 NM_019107.3 Refseq
Affymetrix Human Clariom D Assay PSR1900174771.hg.1 TC1900009357.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900174771.hg.1 ENST00000262947.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900174771.hg.1 ENST00000599630.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900174771.hg.1 ENST00000599761.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900174771.hg.1 NM_019107.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline