This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00964415_m1
See other MOB1A GE Assays ›
Gene Symbol:
MOB1A
Gene Name:
MOB kinase activator 1A
Gene Aliases:
C2orf6, MATS1, MOB1, MOBK1B, MOBKL1B, Mob4B
Chromosome Location:
Chr.2: 74152598 - 74178967 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00964415_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOB1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317110.1 NP_001304039.1 2 - 3 471 110
NM_001317111.1 NP_001304040.1 2 - 3 367 110
NM_001317112.1 NP_001304041.1 2 - 3 474 110
NM_018221.4 NP_060691.2 2 - 3 474 110
XM_011532948.2 XP_011531250.1 2 - 3 334 110
GenBank mRNA AB016839.1 - 2 - 3 323 110
AJ577474.1 - 1 - 2 181 110
AK001650.1 - 2 - 3 364 110
AK021657.1 - 2 - 3 388 110
AK023321.1 - 2 - 3 334 110
BC003398.1 - 2 - 3 375 3448503 110
BI825399.1 - 2 - 3 367 110
BU172616.1 - 2 - 3 370 110
CN426611.1 - 2 - 3 362 110

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200013096.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001993.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MOB1A

Panther Classification:

Molecular Function -

kinase activator

Gene Ontology Categories:

Function(s) Process(es)

hippo signaling
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02031730.hg.1 TC02001993.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02031730.hg.1 ENST00000396049.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02031730.hg.1 ENST00000409969.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02031730.hg.1 ENST00000497054.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02031730.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay PSR0200195592.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200195592.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200195592.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200195592.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200195592.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay PSR0200195592.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200195592.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200195592.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200195592.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200195592.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay PSR0200195592.hg.1 TC0200013096.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0