This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00964426_m1
See other PDGFA GE Assays ›
Gene Symbol:
PDGFA
Gene Name:
platelet derived growth factor subunit A
Gene Aliases:
PDGF-A, PDGF1
Chromosome Location:
Chr.7: 497245 - 520668 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00964426_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer:
284.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PDGFA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002607.5 NP_002598.4 1 - 2 909 67
NM_033023.4 NP_148983.1 1 - 2 909 67
XM_011515415.1 XP_011513717.1 2 - 3 405 67
XM_011515416.1 XP_011513718.1 2 - 3 405 67
XM_011515417.1 XP_011513719.1 2 - 3 405 67
XM_011515418.1 XP_011513720.1 2 - 3 405 67
GenBank mRNA A09204.1 - 1 - 2 102 67
AB451309.1 - 1 - 2 66 67
AB451439.1 - 1 - 2 66 67
AK292217.1 - 1 - 2 280 67
AU118732.1 - 1 - 2 237 67
X03795.1 - 1 - 2 453 67
X06374.1 - 1 - 2 469 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700009977.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07001073.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PDGFA

Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

MAPK cascade
angiogenesis
response to hypoxia
cell activation
hair follicle development
positive regulation of mesenchymal cell proliferation
platelet degranulation
transforming growth factor beta receptor signaling pathway
cell-cell signaling
positive regulation of cell proliferation
response to wounding
organ morphogenesis
response to inorganic substance
negative regulation of phosphatidylinositol biosynthetic process
negative regulation of platelet activation
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of phosphatidylinositol 3-kinase signaling
regulation of smooth muscle cell migration
cell projection assembly
actin cytoskeleton organization
extracellular matrix organization
positive regulation of cell migration
positive regulation of protein autophosphorylation
response to estradiol
response to retinoic acid
regulation of actin cytoskeleton organization
positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway
wound healing
response to drug
positive regulation of MAP kinase activity
positive regulation of MAPK cascade
positive regulation of GTPase activity
skin development
positive regulation of DNA replication
phosphatidylinositol phosphorylation
platelet-derived growth factor receptor signaling pathway
phosphatidylinositol-mediated signaling
positive regulation of fibroblast proliferation
lung alveolus development
inner ear development
regulation of peptidyl-tyrosine phosphorylation
negative chemotaxis
positive regulation of cell division
positive regulation of protein kinase B signaling
regulation of branching involved in salivary gland morphogenesis by epithelial-mesenchymal signaling
positive regulation of ERK1 and ERK2 cascade
regulation of glomerular mesangial cell proliferation
embryonic lung development
regulation of DNA biosynthetic process
Ras guanyl-nucleotide exchange factor activity
platelet-derived growth factor receptor binding
protein binding
collagen binding
growth factor activity
protein homodimerization activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
protein heterodimerization activity
platelet-derived growth factor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07016435.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07016435.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07016435.hg.1 TC07001073.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07016432.hg.1 ENST00000354513.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07016432.hg.1 ENST00000402802.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07016432.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07016432.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07016432.hg.1 TC07001073.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000400761.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000400761.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000400761.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000400761.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000400761.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000400761.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000426681.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000426681.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000482462.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000482462.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000482462.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000482462.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 ENST00000482462.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07016431.hg.1 ENST00000354513.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07016431.hg.1 ENST00000402802.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07016431.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07016431.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07016431.hg.1 TC07001073.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07016429.hg.1 ENST00000354513.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07016429.hg.1 ENST00000402802.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07016429.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07016429.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07016429.hg.1 TC07001073.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07016433.hg.1 ENST00000354513.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07016433.hg.1 ENST00000402802.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07016433.hg.1 NM_002607.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07016433.hg.1 NM_033023.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07016433.hg.1 TC07001073.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0700170979.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay PSR0700170979.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700170979.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700170979.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay PSR0700170979.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay PSR0700170979.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay PSR0700170979.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700170979.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700170979.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay PSR0700170979.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay PSR0700170979.hg.1 TC0700009977.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700170981.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay PSR0700170981.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700170981.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay PSR0700170981.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay PSR0700170981.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay PSR0700170981.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700170981.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay PSR0700170981.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay PSR0700170981.hg.1 TC0700009977.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000400761.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000400761.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000400761.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000400761.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000400761.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000400761.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000426681.2 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000426681.2 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000482462.1 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000482462.1 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000482462.1 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000482462.1 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 ENST00000482462.1 Ensembl
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay TC0700009977.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay PSR0700170983.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay PSR0700170983.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay PSR0700170983.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay PSR0700170983.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay PSR0700170983.hg.1 TC0700009977.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700170977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay PSR0700170977.hg.1 ENST00000400761.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700170977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700170977.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700170977.hg.1 ENST00000426681.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700170977.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay PSR0700170977.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay PSR0700170977.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay PSR0700170977.hg.1 ENST00000400761.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700170977.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700170977.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700170977.hg.1 ENST00000426681.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700170977.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay PSR0700170977.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay PSR0700170977.hg.1 TC0700009977.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700170980.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay PSR0700170980.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700170980.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700170980.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay PSR0700170980.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay PSR0700170980.hg.1 ENST00000354513.9 Ensembl
Affymetrix Human Clariom D Assay PSR0700170980.hg.1 ENST00000402802.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700170980.hg.1 ENST00000405692.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700170980.hg.1 NM_002607.5 Refseq
Affymetrix Human Clariom D Assay PSR0700170980.hg.1 NM_033023.4 Refseq
Affymetrix Human Clariom D Assay PSR0700170980.hg.1 TC0700009977.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline