This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00965855_g1
See other GPSM3 GE Assays ›
Gene Symbol:
GPSM3
Gene Name:
G-protein signaling modulator 3
Gene Aliases:
AGS4, C6orf9, G18, G18.1a, G18.1b, G18.2, NG1
Chromosome Location:
Chr.6: 32190766 - 32195523 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00965855_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPSM3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001276501.1 NP_001263430.1 1 - 2 443 62
NM_022107.2 NP_071390.1 5 - 6 414 62
GenBank mRNA AF155657.1 - 1 - 2 62 62
AJ243937.1 - 5 - 6 414 62
AJ243938.1 - 1 - 2 434 62
AK313922.1 - 1 - 2 443 62
BC018724.1 - 1 - 2 106 4576337 62
DA979033.1 - 1 - 2 443 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600014270.hg.1
This assay targets the same location as HTA transcript cluster(s) TC06001563.hg.1, TC6_apd_hap1000097.hg.1, TC6_cox_hap2000189.hg.1, TC6_mann_hap4000154.hg.1, TC6_mcf_hap5000164.hg.1, TC6_qbl_hap6000178.hg.1, TC6_ssto_hap7000158.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GPSM3

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of leukocyte chemotaxis
biological_process
regulation of catalytic activity
positive regulation of cytokine production involved in inflammatory response
molecular_function
protein binding
GTPase regulator activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5003365.hg.1 TC6_mcf_hap5000164.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5003365.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003518.hg.1 TC6_qbl_hap6000178.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003518.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1001564.hg.1 TC6_apd_hap1000097.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1001564.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003526.hg.1 TC6_qbl_hap6000178.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003526.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7003251.hg.1 TC6_ssto_hap7000158.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7003251.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7003257.hg.1 TC6_ssto_hap7000158.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7003257.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003685.hg.1 TC6_cox_hap2000189.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003685.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002820.hg.1 TC6_mann_hap4000154.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002820.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5003359.hg.1 TC6_mcf_hap5000164.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5003359.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003520.hg.1 TC6_qbl_hap6000178.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_qbl_hap6003520.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1001556.hg.1 TC6_apd_hap1000097.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1001556.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003683.hg.1 TC6_cox_hap2000189.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003683.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7003249.hg.1 TC6_ssto_hap7000158.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_ssto_hap7003249.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002828.hg.1 TC6_mann_hap4000154.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002828.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003691.hg.1 TC6_cox_hap2000189.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_cox_hap2003691.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06018460.hg.1 TC06001563.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06018460.hg.1 ENST00000375040.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06018460.hg.1 ENST00000375043.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06018460.hg.1 ENST00000487761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06018460.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06018468.hg.1 TC06001563.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06018468.hg.1 ENST00000375043.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06018468.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1001558.hg.1 TC6_apd_hap1000097.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_apd_hap1001558.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002822.hg.1 TC6_mann_hap4000154.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_mann_hap4002822.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR06018462.hg.1 TC06001563.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR06018462.hg.1 ENST00000375040.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06018462.hg.1 ENST00000375043.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06018462.hg.1 ENST00000487761.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR06018462.hg.1 NM_022107.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5003357.hg.1 TC6_mcf_hap5000164.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR6_mcf_hap5003357.hg.1 NM_022107.2 Refseq
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Clariom D Assay TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay TC0600014270.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay PSR0600201393.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600201393.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201393.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201393.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay PSR0600201393.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600201393.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201393.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201393.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay PSR0600201393.hg.1 TC0600014270.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600201388.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600201388.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201388.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201388.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600201388.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay PSR0600201388.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600201388.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201388.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201388.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600201388.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay PSR0600201388.hg.1 TC0600014270.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600201390.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600201390.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201390.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201390.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600201390.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay PSR0600201390.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600201390.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201390.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201390.hg.1 ENST00000619085.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600201390.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay PSR0600201390.hg.1 TC0600014270.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600201392.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600201392.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201392.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201392.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay PSR0600201392.hg.1 ENST00000375040.7 Ensembl
Affymetrix Human Clariom D Assay PSR0600201392.hg.1 ENST00000472768.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201392.hg.1 ENST00000487761.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600201392.hg.1 NM_001276501.1 Refseq
Affymetrix Human Clariom D Assay PSR0600201392.hg.1 TC0600014270.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jrd88:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0