This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00966041_m1
See other METTL8 GE Assays ›
Gene Symbol:
METTL8
Gene Name:
methyltransferase like 8
Gene Aliases:
TIP
Chromosome Location:
Chr.2: 171315746 - 171434803 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00966041_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METTL8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024770.4 NP_079046.2 1 - 2 131 100
XM_017004986.1 XP_016860475.1 1 - 2 108 100
GenBank mRNA AK292108.1 - 1 - 2 100 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200014899.hg
This assay targets the same location as HTA transcript cluster(s) TC02002522.hg

Additional Information:

This assay is in the 5'UTR region of the AK292108.1,NM_024770.4,XM_017004986.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene METTL8

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

methylation
methyltransferase activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02040060.hg.1 NM_024770.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02040060.hg.1 TC02002522.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02040050.hg.1 ENST00000375258.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02040050.hg.1 NM_024770.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02040050.hg.1 TC02002522.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000375258.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000392599.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000392604.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000442541.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000442778.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000453846.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000460539.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000612742.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 NM_024770.4 Refseq
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000375258.8 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000392599.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000392604.6 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000442541.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000442778.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000453846.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000460539.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 ENST00000612742.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 NM_024770.4 Refseq
Affymetrix Human Clariom D Assay PSR0200208964.hg.1 TC0200014899.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0200208975.hg.1 ENST00000442778.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200208975.hg.1 ENST00000453846.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200208975.hg.1 ENST00000460188.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200208975.hg.1 ENST00000462821.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200208975.hg.1 ENST00000612742.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200208975.hg.1 NM_024770.4 Refseq
Affymetrix Human Clariom D Assay PSR0200208975.hg.1 ENST00000442778.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200208975.hg.1 ENST00000453846.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200208975.hg.1 ENST00000460188.5 Ensembl
Affymetrix Human Clariom D Assay PSR0200208975.hg.1 ENST00000462821.1 Ensembl
Affymetrix Human Clariom D Assay PSR0200208975.hg.1 ENST00000612742.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200208975.hg.1 NM_024770.4 Refseq
Affymetrix Human Clariom D Assay PSR0200208975.hg.1 TC0200014899.hg.1 Clariom D TC
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 222982 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline