This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00966048_g1
See other METTL8 GE Assays ›
Gene Symbol:
METTL8
Gene Name:
methyltransferase like 8
Gene Aliases:
TIP
Chromosome Location:
Chr.2: 171315746 - 171434803 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00966048_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METTL8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321154.1 NP_001308083.1 8 - 9 1200 135
NM_001321155.1 NP_001308084.1 9 - 10 1324 135
NM_001321156.1 NP_001308085.1 9 - 10 1394 135
NM_001321157.1 NP_001308086.1 9 - 10 1227 135
NM_001321158.1 NP_001308087.1 7 - 8 948 135
NM_001321159.1 NP_001308088.1 7 - 8 1045 135
NM_024770.4 NP_079046.2 8 - 9 1103 135
NR_135568.1 5 - 6 445 135
XM_017004984.1 XP_016860473.1 9 - 10 1243 135
XM_017004985.1 XP_016860474.1 9 - 10 1341 135
XM_017004986.1 XP_016860475.1 8 - 9 1119 135
XM_017004987.1 XP_016860476.1 7 - 8 981 135
XM_017004988.1 XP_016860477.1 7 - 8 965 135
XM_017004989.1 XP_016860478.1 8 - 9 1134 135
GenBank mRNA AK093218.1 - 3 - 4 1233 135
AK124092.1 - 9 - 10 1275 135
AK292108.1 - 8 - 9 1072 135
AK297141.1 - 7 - 8 948 135
DB031573.1 - 5 - 6 430 135

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC02002522.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene METTL8

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

methylation
methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR02040028.hg.1 ENST00000375258.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02040028.hg.1 ENST00000463392.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02040028.hg.1 NM_024770.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02040028.hg.1 TC02002522.hg.1 HTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline