This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00966722_m1
See other RALGAPB GE Assays ›
Gene Symbol:
RALGAPB
Gene Name:
Ral GTPase activating protein non-catalytic beta subunit
Gene Aliases:
KIAA1219, RalGAPbeta
Chromosome Location:
Chr.20: 38472806 - 38578861 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00966722_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RALGAPB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282917.1 NP_001269846.1 24 - 25 3853 118
NM_020336.3 NP_065069.1 24 - 25 3995 118
XM_005260463.2 XP_005260520.1 25 - 26 3989 118
XM_017027966.1 XP_016883455.1 25 - 26 3977 118
XM_017027967.1 XP_016883456.1 24 - 25 3941 118
XM_017027969.1 XP_016883458.1 25 - 26 4006 118
GenBank mRNA BC018668.1 - 3 - 4 488 118
BC047482.1 - 24 - 25 3958 5270777 118
CR936800.1 - 24 - 25 3759 118

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2000007320.hg.1
This assay targets the same location as HTA transcript cluster(s) TC20000285.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RALGAPB

Gene Ontology Categories:

Function(s) Process(es)

regulation of small GTPase mediated signal transduction
activation of GTPase activity
GTPase activator activity
protein heterodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR20004109.hg.1 ENST00000262879.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20004109.hg.1 ENST00000397038.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20004109.hg.1 ENST00000397040.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20004109.hg.1 ENST00000397042.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR20004109.hg.1 NM_020336.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR20004109.hg.1 TC20000285.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 ENST00000262879.10 Ensembl
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 ENST00000397040.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 ENST00000397042.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 ENST00000438490.2 Ensembl
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 ENST00000632792.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 NM_001282917.1 Refseq
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 NM_001282918.1 Refseq
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 NM_020336.3 Refseq
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 TC2000007320.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 ENST00000262879.10 Ensembl
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 ENST00000397040.5 Ensembl
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 ENST00000397042.7 Ensembl
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 ENST00000438490.2 Ensembl
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 ENST00000632792.1 Ensembl
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 NM_001282917.1 Refseq
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 NM_001282918.1 Refseq
Affymetrix Human Clariom D Assay PSR2000150688.hg.1 NM_020336.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0