This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00968250_m1
See other MOB1A GE Assays ›
Gene Symbol:
MOB1A
Gene Name:
MOB kinase activator 1A
Gene Aliases:
C2orf6, MATS1, MOB1, MOBK1B, MOBKL1B, Mob4B
Chromosome Location:
Chr.2: 74152598 - 74178967 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00968250_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOB1A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001317110.1 NP_001304039.1 4 - 5 701 158
NM_001317111.1 NP_001304040.1 4 - 5 597 158
NM_018221.4 NP_060691.2 4 - 5 704 158
GenBank mRNA AB016839.1 - 4 - 5 553 158
AJ577474.1 - 3 - 4 411 158
AK001650.1 - 4 - 5 594 158
AK023321.1 - 4 - 5 564 158
BC003398.1 - 4 - 5 605 3448503 158
BI825399.1 - 4 - 5 597 158

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0200013096.hg.1
This assay targets the same location as HTA transcript cluster(s) TC02001993.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MOB1A

Panther Classification:

Molecular Function -

kinase activator

Gene Ontology Categories:

Function(s) Process(es)

hippo signaling
protein binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR02031725.hg.1 TC02001993.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR02031725.hg.1 ENST00000396049.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR02031725.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay PSR0200195587.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200195587.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay PSR0200195587.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay PSR0200195587.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay PSR0200195587.hg.1 TC0200013096.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000396049.4 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000463975.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000488006.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000494600.1 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000495286.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 ENST00000497054.5 Ensembl
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq
Affymetrix Human Clariom D Assay TC0200013096.hg.1 NM_018221.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0