This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00969197_g1
See other METTL26 GE Assays ›
Gene Symbol:
METTL26
Gene Name:
methyltransferase like 26
Gene Aliases:
C16orf13, JFP2
Chromosome Location:
Chr.16: 634427 - 636366 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00969197_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METTL26
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001288710.1 NP_001275639.1 3 - 4 506 122
NM_032366.4 NP_115742.3 4 - 5 566 122
NR_109976.1 3 - 4 510 122
NR_109977.1 3 - 4 403 122
NR_109978.1 4 - 5 673 122
NR_109979.1 2 - 3 343 122
XM_011522713.2 XP_011521015.1 4 - 5 765 122
GenBank mRNA AK057077.1 - 1 - 2 1464 122
AK294482.1 - 4 - 5 502 122
AW027232.1 - 3 - 4 422 122
BG612714.1 - 3 - 4 367 122
BI762987.1 - 4 - 5 542 122
BP273946.1 - 3 - 4 442 122
BQ921757.1 - 471 122
BU539308.1 - 2 - 3 291 122
BX104467.1 - 2 - 3 122

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene METTL26

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0