This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00969319_m1
See other NEDD4L GE Assays ›
Gene Symbol:
NEDD4L
Gene Name:
neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase
Gene Aliases:
NEDD4-2, NEDD4.2, RSP5, hNEDD4-2
Chromosome Location:
Chr.18: 58044362 - 58401540 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00969319_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NEDD4L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144964.1 NP_001138436.1 15 - 16 1615 63
NM_001144965.1 NP_001138437.1 15 - 16 1567 63
NM_001144966.2 NP_001138438.1 14 - 15 1492 63
NM_001144967.2 NP_001138439.1 15 - 16 1661 63
NM_001144968.1 NP_001138440.1 15 - 16 1557 63
NM_001144969.1 NP_001138441.1 14 - 15 1497 63
NM_001144970.2 NP_001138442.1 13 - 14 1540 63
NM_001144971.1 NP_001138443.1 15 - 16 1585 63
NM_001243960.1 NP_001230889.1 13 - 14 1469 63
NM_015277.5 NP_056092.2 14 - 15 1601 63
XM_005266658.4 XP_005266715.2 11 - 12 1538 63
XM_005266660.4 XP_005266717.2 11 - 12 1536 63
XM_005266663.4 XP_005266720.2 10 - 11 1476 63
XM_006722421.3 XP_006722484.2 10 - 11 1477 63
XM_006722426.3 XP_006722489.1 16 - 17 1755 63
XM_006722428.3 XP_006722491.1 15 - 16 1677 63
XM_006722430.3 XP_006722493.1 14 - 15 1585 63
XM_011525887.2 XP_011524189.1 16 - 17 1735 63
XM_017025678.1 XP_016881167.1 14 - 15 1616 63
XM_017025679.1 XP_016881168.1 14 - 15 1585 63
GenBank mRNA AB007899.1 - 13 - 14 1440 63
AB071179.1 - 14 - 15 1432 63
AF210730.1 - 14 - 15 1392 63
AF385931.1 - 15 - 16 1616 63
AK294963.1 - 13 - 14 1489 63
AK300444.1 - 14 - 15 1443 63
AY112983.1 - 15 - 16 1567 63
AY112984.2 - 15 - 16 1557 63
AY112985.2 - 14 - 15 1501 63
AY312514.1 - 15 - 16 1378 63
BC000621.2 - 9 - 10 1018 63
BC019345.1 - 9 - 10 1018 63
BC032597.1 - 13 - 14 1496 5528964 63
DQ181796.1 - 15 - 16 1585 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800007411.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000202.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NEDD4L

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
protein polyubiquitination
regulation of membrane depolarization
protein monoubiquitination
sodium ion transport
cellular sodium ion homeostasis
excretion
response to metal ion
protein ubiquitination
viral life cycle
water homeostasis
ion transmembrane transport
regulation of ion transmembrane transport
regulation of protein catabolic process
regulation of membrane potential
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of protein catabolic process
positive regulation of endocytosis
regulation of membrane repolarization
protein K48-linked ubiquitination
ventricular cardiac muscle cell action potential
regulation of potassium ion transmembrane transporter activity
negative regulation of potassium ion transmembrane transporter activity
negative regulation of potassium ion transmembrane transport
negative regulation of sodium ion transmembrane transport
positive regulation of dendrite extension
negative regulation of protein localization to cell surface
negative regulation of sodium ion transmembrane transporter activity
positive regulation of caveolin-mediated endocytosis
ubiquitin-protein transferase activity
protein binding
potassium channel regulator activity
ligase activity
sodium channel regulator activity
potassium channel inhibitor activity
sodium channel inhibitor activity
ion channel binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0