This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00969331_m1
See other NEDD4L GE Assays ›
Gene Symbol:
NEDD4L
Gene Name:
neural precursor cell expressed, developmentally down-regulated 4-like, E3 ubiquitin protein ligase
Gene Aliases:
NEDD4-2, NEDD4.2, RSP5, hNEDD4-2
Chromosome Location:
Chr.18: 58044362 - 58401540 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00969331_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NEDD4L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001144964.1 NP_001138436.1 26 - 27 2721 97
NM_001144965.1 NP_001138437.1 26 - 27 2673 97
NM_001144966.2 NP_001138438.1 25 - 26 2598 97
NM_001144967.2 NP_001138439.1 26 - 27 2767 97
NM_001144968.1 NP_001138440.1 26 - 27 2663 97
NM_001144969.1 NP_001138441.1 25 - 26 2603 97
NM_001144970.2 NP_001138442.1 24 - 25 2646 97
NM_001144971.1 NP_001138443.1 26 - 27 2691 97
NM_001243960.1 NP_001230889.1 24 - 25 2575 97
NM_015277.5 NP_056092.2 25 - 26 2707 97
XM_005266658.4 XP_005266715.2 23 - 24 2698 97
XM_005266660.4 XP_005266717.2 22 - 23 2642 97
XM_005266663.4 XP_005266720.2 21 - 22 2582 97
XM_006722421.3 XP_006722484.2 22 - 23 2637 97
XM_006722424.3 XP_006722487.2 19 - 20 2190 97
XM_006722425.3 XP_006722488.2 18 - 19 2134 97
XM_006722426.3 XP_006722489.1 28 - 29 2915 97
XM_006722428.3 XP_006722491.1 27 - 28 2837 97
XM_006722430.3 XP_006722493.1 26 - 27 2745 97
XM_011525887.2 XP_011524189.1 28 - 29 2895 97
XM_017025676.1 XP_016881165.1 20 - 21 2387 97
XM_017025677.1 XP_016881166.1 19 - 20 2331 97
XM_017025678.1 XP_016881167.1 26 - 27 2776 97
XM_017025679.1 XP_016881168.1 25 - 26 2691 97
XM_017025680.1 XP_016881169.1 22 - 23 2379 97
XM_017025681.1 XP_016881170.1 21 - 22 2181 97
GenBank mRNA AK294963.1 - 25 - 26 2598 97
AK300444.1 - 25 - 26 2549 97
AL137469.1 - 21 - 22 2019 97
AY112983.1 - 26 - 27 2673 97
AY112984.2 - 26 - 27 2663 97
BC000621.2 - 20 - 21 2124 97
BC019345.1 - 20 - 21 2124 97
BC032597.1 - 24 - 25 2602 5528964 97
BQ215431.1 - 5 - 6 331 97
DQ181796.1 - 26 - 27 2691 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800007411.hg.1
This assay targets the same location as HTA transcript cluster(s) TC18000202.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NEDD4L

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
protein polyubiquitination
regulation of membrane depolarization
protein monoubiquitination
sodium ion transport
cellular sodium ion homeostasis
excretion
response to metal ion
protein ubiquitination
viral life cycle
water homeostasis
ion transmembrane transport
regulation of ion transmembrane transport
regulation of protein catabolic process
regulation of membrane potential
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
positive regulation of protein catabolic process
positive regulation of endocytosis
regulation of membrane repolarization
protein K48-linked ubiquitination
ventricular cardiac muscle cell action potential
regulation of potassium ion transmembrane transporter activity
negative regulation of potassium ion transmembrane transporter activity
negative regulation of potassium ion transmembrane transport
negative regulation of sodium ion transmembrane transport
positive regulation of dendrite extension
negative regulation of protein localization to cell surface
negative regulation of sodium ion transmembrane transporter activity
positive regulation of caveolin-mediated endocytosis
ubiquitin-protein transferase activity
protein binding
potassium channel regulator activity
ligase activity
sodium channel regulator activity
potassium channel inhibitor activity
sodium channel inhibitor activity
ion channel binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0